Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 149350717-149350978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRPEL2
Entrez Gene ID
134266
Gene Name
GrpE like 2, mitochondrial
Gene Aliases
Mt-GrpE#2
UniGene
Hs.511816
Gene Chromosome Location
Chr.5: 149345414-149354583 on Build GRCh38
Transcripts
NM_152407.3 NR_132366.1

Gene Symbol
GRPEL2-AS1
Entrez Gene ID
106144529
Gene Name
GRPEL2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.607162
Gene Chromosome Location
Chr.5: 149348116-149357642 on Build GRCh38
Transcripts
NM_152407.3 NR_132366.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3994108 c.327C>T p.F109F Chr.5: 149350931-149350931 on Build GRCh38
3947123 c.352G>T p.D118Y Chr.5: 149350956-149350956 on Build GRCh38
7252582 c.352G>C p.D118H Chr.5: 149350956-149350956 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766829733 Chr.5: 149350897-149350897 on Build GRCh38
rs1045297116 Chr.5: 149350806-149350806 on Build GRCh38
rs139055625 Chr.5: 149350929-149350929 on Build GRCh38
rs753197148 Chr.5: 149350952-149350952 on Build GRCh38
rs532415277 Chr.5: 149350860-149350860 on Build GRCh38
rs1024505337 Chr.5: 149350931-149350931 on Build GRCh38
rs776270650 Chr.5: 149350867-149350867 on Build GRCh38
rs765260914 Chr.5: 149350829-149350829 on Build GRCh38
rs747878548 Chr.5: 149350868-149350868 on Build GRCh38
rs184666720 Chr.5: 149350941-149350941 on Build GRCh38
rs898723781 Chr.5: 149350821-149350821 on Build GRCh38
rs752704936 Chr.5: 149350947-149350947 on Build GRCh38
rs763332812 Chr.5: 149350893-149350893 on Build GRCh38
rs948318747 Chr.5: 149350803-149350803 on Build GRCh38
rs774967016 Chr.5: 149350910-149350910 on Build GRCh38
rs1013328943 Chr.5: 149350908-149350908 on Build GRCh38
rs866280701 Chr.5: 149350779-149350779 on Build GRCh38
rs773465841 Chr.5: 149350872-149350872 on Build GRCh38
rs781779107 Chr.5: 149350942-149350944 on Build GRCh38
rs937681543 Chr.5: 149350838-149350838 on Build GRCh38
rs913731511 Chr.5: 149350744-149350744 on Build GRCh38
rs945325246 Chr.5: 149350761-149350761 on Build GRCh38
rs760018184 Chr.5: 149350928-149350928 on Build GRCh38
rs769508156 Chr.5: 149350870-149350870 on Build GRCh38
rs757683030 Chr.5: 149350886-149350889 on Build GRCh38

View Full Product Details