Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24336699-24336924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
UniGene
Hs.657920
Gene Chromosome Location
Chr.1: 24319322-24364482 on Build GRCh38
Transcripts
NM_001195010.1 NM_021180.3 NM_198173.2 NM_198174.2 XM_011541869.1 XM_011541870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2149734 c.512C>T p.P171L Chr.1: 24336712-24336712 on Build GRCh38
4688477 c.560C>T p.P187L Chr.1: 24336775-24336775 on Build GRCh38
4552366 c.556G>C p.E186Q Chr.1: 24336756-24336756 on Build GRCh38
4030440 c.586C>T p.R196C Chr.1: 24336786-24336786 on Build GRCh38
4751356 c.546C>G p.N182K Chr.1: 24336746-24336746 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770785729 Chr.1: 24336823-24336823 on Build GRCh38
rs759002627 Chr.1: 24336864-24336864 on Build GRCh38
rs368857465 Chr.1: 24336750-24336750 on Build GRCh38
rs191739265 Chr.1: 24336767-24336767 on Build GRCh38
rs201895291 Chr.1: 24336836-24336836 on Build GRCh38
rs775035732 Chr.1: 24336840-24336840 on Build GRCh38
rs762034246 Chr.1: 24336859-24336859 on Build GRCh38
rs147556033 Chr.1: 24336852-24336852 on Build GRCh38
rs373482025 Chr.1: 24336753-24336753 on Build GRCh38
rs763774678 Chr.1: 24336848-24336848 on Build GRCh38
rs984626770 Chr.1: 24336815-24336815 on Build GRCh38
rs947657260 Chr.1: 24336884-24336884 on Build GRCh38
rs777682242 Chr.1: 24336810-24336810 on Build GRCh38
rs774380464 Chr.1: 24336724-24336724 on Build GRCh38
rs144050104 Chr.1: 24336728-24336728 on Build GRCh38
rs779246351 Chr.1: 24336761-24336761 on Build GRCh38
rs752284034 Chr.1: 24336731-24336731 on Build GRCh38
rs765647968 Chr.1: 24336862-24336862 on Build GRCh38
rs112149898 Chr.1: 24336774-24336774 on Build GRCh38
rs964530884 Chr.1: 24336833-24336833 on Build GRCh38
rs751730977 Chr.1: 24336875-24336875 on Build GRCh38
rs923030066 Chr.1: 24336844-24336844 on Build GRCh38
rs144957254 Chr.1: 24336821-24336821 on Build GRCh38
rs116674530 Chr.1: 24336775-24336775 on Build GRCh38
rs150060454 Chr.1: 24336787-24336787 on Build GRCh38
rs780260695 Chr.1: 24336867-24336867 on Build GRCh38
rs758924862 Chr.1: 24336725-24336725 on Build GRCh38
rs771474229 Chr.1: 24336839-24336839 on Build GRCh38
rs760295976 Chr.1: 24336843-24336843 on Build GRCh38
rs755826508 Chr.1: 24336736-24336736 on Build GRCh38
rs187353560 Chr.1: 24336900-24336900 on Build GRCh38
rs774101755 Chr.1: 24336835-24336835 on Build GRCh38
rs200674358 Chr.1: 24336786-24336786 on Build GRCh38
rs145763021 Chr.1: 24336890-24336891 on Build GRCh38
rs780285232 Chr.1: 24336776-24336776 on Build GRCh38
rs757749100 Chr.1: 24336755-24336755 on Build GRCh38
rs35868286 Chr.1: 24336722-24336722 on Build GRCh38

View Full Product Details