Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 69232565-69232826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HKDC1
Entrez Gene ID
80201
Gene Name
hexokinase domain containing 1
Gene Aliases
-
UniGene
Hs.522988
Gene Chromosome Location
Chr.10: 69220303-69267559 on Build GRCh38
Transcripts
NM_025130.3 XM_011540195.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5831500 c.258_259insG p.S88fs*12 Chr.10: 69232795-69232796 on Build GRCh38
5907546 c.244T>G p.S82A Chr.10: 69232781-69232781 on Build GRCh38
7209896 c.238T>C p.F80L Chr.10: 69232775-69232775 on Build GRCh38
4480844 c.245C>T p.S82F Chr.10: 69232782-69232782 on Build GRCh38
7209895 c.238T>C p.F80L Chr.10: 69232775-69232775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573020810 Chr.10: 69232675-69232675 on Build GRCh38
rs909488440 Chr.10: 69232721-69232721 on Build GRCh38
rs576020521 Chr.10: 69232735-69232735 on Build GRCh38
rs758640909 Chr.10: 69232792-69232792 on Build GRCh38
rs963809354 Chr.10: 69232613-69232613 on Build GRCh38
rs756419352 Chr.10: 69232588-69232588 on Build GRCh38
rs1005150079 Chr.10: 69232596-69232596 on Build GRCh38
rs992296805 Chr.10: 69232660-69232660 on Build GRCh38
rs777175998 Chr.10: 69232744-69232744 on Build GRCh38
rs749894632 Chr.10: 69232749-69232750 on Build GRCh38
rs189831527 Chr.10: 69232676-69232676 on Build GRCh38
rs150293872 Chr.10: 69232646-69232646 on Build GRCh38
rs947121299 Chr.10: 69232751-69232751 on Build GRCh38
rs762084273 Chr.10: 69232628-69232628 on Build GRCh38
rs539517093 Chr.10: 69232680-69232680 on Build GRCh38
rs765568117 Chr.10: 69232756-69232756 on Build GRCh38
rs1051670927 Chr.10: 69232798-69232798 on Build GRCh38
rs140271381 Chr.10: 69232774-69232774 on Build GRCh38
rs374874844 Chr.10: 69232808-69232808 on Build GRCh38
rs762144777 Chr.10: 69232752-69232752 on Build GRCh38
rs1016326675 Chr.10: 69232598-69232598 on Build GRCh38
rs767692707 Chr.10: 69232638-69232638 on Build GRCh38
rs764629616 Chr.10: 69232727-69232727 on Build GRCh38
rs874557 Chr.10: 69232657-69232657 on Build GRCh38
rs372141992 Chr.10: 69232778-69232778 on Build GRCh38
rs1022227242 Chr.10: 69232629-69232629 on Build GRCh38
rs1038840704 Chr.10: 69232769-69232769 on Build GRCh38
rs1033711015 Chr.10: 69232654-69232654 on Build GRCh38
rs982316065 Chr.10: 69232647-69232647 on Build GRCh38
rs201928386 Chr.10: 69232793-69232793 on Build GRCh38
rs984970958 Chr.10: 69232683-69232683 on Build GRCh38
rs761205637 Chr.10: 69232725-69232725 on Build GRCh38

View Full Product Details