Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 16152451-16152722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSH2D
Entrez Gene ID
84941
Gene Name
hematopoietic SH2 domain containing
Gene Aliases
ALX, HSH2
UniGene
Hs.631617
Gene Chromosome Location
Chr.19: 16134028-16158575 on Build GRCh38
Transcripts
NM_001291274.1 NM_032855.3 NR_111903.1 NR_111904.1 XM_011528386.2 XM_011528387.2 XM_011528388.2 XM_011528389.2 XM_011528390.1 XM_017027402.1 XM_017027403.1 XM_017027404.1

SNP Detail

SNP ID Chromosome Location
rs765647615 Chr.19: 16152699-16152699 on Build GRCh38
rs771116890 Chr.19: 16152697-16152697 on Build GRCh38
rs368091372 Chr.19: 16152519-16152519 on Build GRCh38
rs778889592 Chr.19: 16152654-16152654 on Build GRCh38
rs758240190 Chr.19: 16152672-16152672 on Build GRCh38
rs760337514 Chr.19: 16152627-16152627 on Build GRCh38
rs115528976 Chr.19: 16152510-16152510 on Build GRCh38
rs747224696 Chr.19: 16152678-16152678 on Build GRCh38
rs747994855 Chr.19: 16152536-16152536 on Build GRCh38
rs776731700 Chr.19: 16152561-16152561 on Build GRCh38
rs377308378 Chr.19: 16152650-16152650 on Build GRCh38
rs887987336 Chr.19: 16152491-16152491 on Build GRCh38
rs370423766 Chr.19: 16152503-16152503 on Build GRCh38
rs1037522600 Chr.19: 16152703-16152703 on Build GRCh38
rs750147792 Chr.19: 16152621-16152621 on Build GRCh38
rs778011426 Chr.19: 16152539-16152539 on Build GRCh38
rs891131944 Chr.19: 16152688-16152688 on Build GRCh38
rs754881083 Chr.19: 16152523-16152523 on Build GRCh38
rs368687301 Chr.19: 16152628-16152628 on Build GRCh38
rs759992364 Chr.19: 16152605-16152605 on Build GRCh38
rs770277045 Chr.19: 16152583-16152583 on Build GRCh38
rs776123188 Chr.19: 16152600-16152600 on Build GRCh38
rs747211497 Chr.19: 16152546-16152546 on Build GRCh38
rs372296907 Chr.19: 16152633-16152633 on Build GRCh38
rs1048057527 Chr.19: 16152480-16152480 on Build GRCh38
rs558813360 Chr.19: 16152489-16152489 on Build GRCh38
rs370279304 Chr.19: 16152532-16152532 on Build GRCh38
rs779569579 Chr.19: 16152511-16152511 on Build GRCh38
rs201712455 Chr.19: 16152537-16152537 on Build GRCh38
rs770936437 Chr.19: 16152559-16152559 on Build GRCh38
rs557515540 Chr.19: 16152702-16152702 on Build GRCh38
rs764519789 Chr.19: 16152618-16152618 on Build GRCh38
rs746018916 Chr.19: 16152701-16152701 on Build GRCh38
rs763475192 Chr.19: 16152615-16152615 on Build GRCh38
rs932905784 Chr.19: 16152656-16152656 on Build GRCh38
rs369352376 Chr.19: 16152701-16152702 on Build GRCh38
rs781144370 Chr.19: 16152700-16152700 on Build GRCh38
rs575204510 Chr.19: 16152568-16152568 on Build GRCh38
rs752600857 Chr.19: 16152655-16152655 on Build GRCh38

View Full Product Details