Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30909322-30909583 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H4
Entrez Gene ID
2968
Gene Name
general transcription factor IIH subunit 4
Gene Aliases
P52, TFB2, TFIIH
UniGene
Hs.485070
Gene Chromosome Location
Chr.6: 30908200-30914103 on Build GRCh38
Transcripts
NM_001517.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6676727 c.211G>T p.V71L Chr.6: 30909508-30909508 on Build GRCh38
6431709 c.191A>G p.Q64R Chr.6: 30909488-30909488 on Build GRCh38
6431707 c.191A>G p.Q64R Chr.6: 30909488-30909488 on Build GRCh38
6676729 c.211G>T p.V71L Chr.6: 30909508-30909508 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs142542393 Chr.6: 30909486-30909486 on Build GRCh38
rs766572667 Chr.6: 30909505-30909505 on Build GRCh38
rs79233378 Chr.6: 30909506-30909506 on Build GRCh38
rs762738144 Chr.6: 30909509-30909509 on Build GRCh38
rs916920 Chr.6: 30909425-30909425 on Build GRCh38
rs950936569 Chr.6: 30909397-30909397 on Build GRCh38
rs530452070 Chr.6: 30909423-30909423 on Build GRCh38
rs937489495 Chr.6: 30909344-30909344 on Build GRCh38
rs764184133 Chr.6: 30909474-30909474 on Build GRCh38
rs566998262 Chr.6: 30909445-30909445 on Build GRCh38
rs763833945 Chr.6: 30909511-30909511 on Build GRCh38
rs140677952 Chr.6: 30909537-30909537 on Build GRCh38
rs3869090 Chr.6: 30909481-30909481 on Build GRCh38
rs752970265 Chr.6: 30909374-30909374 on Build GRCh38
rs1024788977 Chr.6: 30909434-30909434 on Build GRCh38
rs150962097 Chr.6: 30909490-30909490 on Build GRCh38
rs749686225 Chr.6: 30909427-30909427 on Build GRCh38
rs3906277 Chr.6: 30909456-30909456 on Build GRCh38
rs1057260274 Chr.6: 30909381-30909381 on Build GRCh38
rs748555174 Chr.6: 30909470-30909470 on Build GRCh38
rs745578291 Chr.6: 30909421-30909421 on Build GRCh38
rs780514850 Chr.6: 30909398-30909398 on Build GRCh38
rs750323550 Chr.6: 30909503-30909503 on Build GRCh38
rs756819978 Chr.6: 30909392-30909392 on Build GRCh38
rs752728407 Chr.6: 30909462-30909462 on Build GRCh38

View Full Product Details