Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119048517-119048742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
UniGene
Hs.277704
Gene Chromosome Location
Chr.11: 119044185-119057246 on Build GRCh38
Transcripts
NM_001130991.2 NM_006389.4 XM_005271392.3 XM_005271393.3 XM_005271394.3 XM_017017095.1 XM_017017096.1 XM_017017097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4965744 c.2166-5C>T p.? Chr.11: 119048568-119048568 on Build GRCh38
7371735 c.2206C>T p.R736W Chr.11: 119048523-119048523 on Build GRCh38
1242362 c.2157G>A p.S719S Chr.11: 119048722-119048722 on Build GRCh38
1254565 c.2156C>T p.S719L Chr.11: 119048723-119048723 on Build GRCh38
7306652 c.2185C>T p.R729* Chr.11: 119048544-119048544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782126475 Chr.11: 119048721-119048721 on Build GRCh38
rs566850822 Chr.11: 119048700-119048700 on Build GRCh38
rs147525277 Chr.11: 119048649-119048649 on Build GRCh38
rs782738483 Chr.11: 119048722-119048722 on Build GRCh38
rs782163982 Chr.11: 119048590-119048590 on Build GRCh38
rs782342968 Chr.11: 119048558-119048558 on Build GRCh38
rs987838110 Chr.11: 119048581-119048581 on Build GRCh38
rs564777944 Chr.11: 119048603-119048603 on Build GRCh38
rs782457061 Chr.11: 119048597-119048597 on Build GRCh38
rs782513791 Chr.11: 119048683-119048694 on Build GRCh38
rs2276059 Chr.11: 119048666-119048666 on Build GRCh38
rs532331360 Chr.11: 119048627-119048627 on Build GRCh38
rs983551572 Chr.11: 119048671-119048671 on Build GRCh38
rs782586020 Chr.11: 119048598-119048598 on Build GRCh38
rs782148656 Chr.11: 119048690-119048690 on Build GRCh38
rs201533554 Chr.11: 119048714-119048714 on Build GRCh38
rs965679428 Chr.11: 119048701-119048701 on Build GRCh38
rs141569642 Chr.11: 119048641-119048641 on Build GRCh38
rs148836750 Chr.11: 119048543-119048543 on Build GRCh38
rs782225234 Chr.11: 119048679-119048679 on Build GRCh38
rs143389635 Chr.11: 119048548-119048548 on Build GRCh38
rs782749634 Chr.11: 119048592-119048592 on Build GRCh38
rs782671293 Chr.11: 119048685-119048686 on Build GRCh38
rs544181749 Chr.11: 119048628-119048628 on Build GRCh38
rs372068451 Chr.11: 119048544-119048544 on Build GRCh38
rs782242670 Chr.11: 119048574-119048576 on Build GRCh38
rs782687466 Chr.11: 119048606-119048606 on Build GRCh38
rs202126223 Chr.11: 119048593-119048593 on Build GRCh38
rs782712882 Chr.11: 119048582-119048582 on Build GRCh38
rs782395503 Chr.11: 119048669-119048669 on Build GRCh38
rs782051651 Chr.11: 119048580-119048580 on Build GRCh38
rs781884275 Chr.11: 119048583-119048583 on Build GRCh38
rs781933792 Chr.11: 119048689-119048689 on Build GRCh38
rs376359962 Chr.11: 119048579-119048579 on Build GRCh38
rs782306849 Chr.11: 119048683-119048683 on Build GRCh38
rs781984097 Chr.11: 119048702-119048702 on Build GRCh38
rs782595910 Chr.11: 119048674-119048674 on Build GRCh38
rs782542840 Chr.11: 119048604-119048604 on Build GRCh38

View Full Product Details