Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72229634-72229867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6850225 c.772G>A p.E258K Chr.11: 72229681-72229681 on Build GRCh38
5492838 c.754-8delC p.? Chr.11: 72229655-72229655 on Build GRCh38
3810362 c.827C>G p.S276* Chr.11: 72229736-72229736 on Build GRCh38
4805222 c.754-2A>G p.? Chr.11: 72229661-72229661 on Build GRCh38
5557883 c.799A>G p.K267E Chr.11: 72229708-72229708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548467739 Chr.11: 72229796-72229796 on Build GRCh38
rs760757952 Chr.11: 72229752-72229752 on Build GRCh38
rs951723805 Chr.11: 72229792-72229792 on Build GRCh38
rs566700152 Chr.11: 72229822-72229822 on Build GRCh38
rs775043885 Chr.11: 72229655-72229655 on Build GRCh38
rs774613122 Chr.11: 72229728-72229728 on Build GRCh38
rs777376401 Chr.11: 72229677-72229677 on Build GRCh38
rs758056866 Chr.11: 72229781-72229781 on Build GRCh38
rs745668155 Chr.11: 72229707-72229707 on Build GRCh38
rs371206323 Chr.11: 72229715-72229715 on Build GRCh38
rs770929582 Chr.11: 72229692-72229692 on Build GRCh38
rs746518063 Chr.11: 72229683-72229683 on Build GRCh38
rs568210147 Chr.11: 72229759-72229759 on Build GRCh38
rs755346094 Chr.11: 72229778-72229778 on Build GRCh38
rs746501297 Chr.11: 72229798-72229798 on Build GRCh38
rs1007477970 Chr.11: 72229828-72229828 on Build GRCh38
rs747802572 Chr.11: 72229671-72229671 on Build GRCh38
rs1003397653 Chr.11: 72229693-72229693 on Build GRCh38
rs778596194 Chr.11: 72229664-72229664 on Build GRCh38
rs767762060 Chr.11: 72229734-72229734 on Build GRCh38
rs539048364 Chr.11: 72229767-72229767 on Build GRCh38
rs887200823 Chr.11: 72229732-72229732 on Build GRCh38
rs80156982 Chr.11: 72229705-72229705 on Build GRCh38
rs776376700 Chr.11: 72229782-72229783 on Build GRCh38
rs201713633 Chr.11: 72229659-72229659 on Build GRCh38
rs797044468 Chr.11: 72229677-72229677 on Build GRCh38
rs61736300 Chr.11: 72229710-72229710 on Build GRCh38
rs529907144 Chr.11: 72229772-72229772 on Build GRCh38
rs527470737 Chr.11: 72229837-72229837 on Build GRCh38
rs900076023 Chr.11: 72229846-72229846 on Build GRCh38
rs374049262 Chr.11: 72229660-72229660 on Build GRCh38
rs761978331 Chr.11: 72229731-72229731 on Build GRCh38
rs776487134 Chr.11: 72229703-72229703 on Build GRCh38
rs1005517470 Chr.11: 72229776-72229776 on Build GRCh38
rs374410580 Chr.11: 72229780-72229780 on Build GRCh38
rs768343273 Chr.11: 72229776-72229776 on Build GRCh38
rs369528817 Chr.11: 72229831-72229831 on Build GRCh38
rs766877002 Chr.11: 72229658-72229658 on Build GRCh38
rs552436243 Chr.11: 72229847-72229847 on Build GRCh38
rs758146582 Chr.11: 72229676-72229676 on Build GRCh38
rs746647683 Chr.11: 72229677-72229678 on Build GRCh38
rs773210890 Chr.11: 72229740-72229740 on Build GRCh38

View Full Product Details