Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156752063-156752336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDGF
Entrez Gene ID
3068
Gene Name
hepatoma-derived growth factor
Gene Aliases
HMG1L2
UniGene
Hs.743948
Gene Chromosome Location
Chr.1: 156742107-156752448 on Build GRCh38
Transcripts
NM_001126050.1 NM_001126051.1 NM_001319186.1 NR_135008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5768356 c.33G>C p.Q11H Chr.1: 156752071-156752071 on Build GRCh38
4633347 c.85G>T p.G29C Chr.1: 156752279-156752279 on Build GRCh38
3356450 c.31C>T p.Q11* Chr.1: 156752073-156752073 on Build GRCh38
6039422 c.130delC p.H44fs*11 Chr.1: 156752234-156752234 on Build GRCh38
897356 c.106C>T p.P36S Chr.1: 156752258-156752258 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750397944 Chr.1: 156752131-156752131 on Build GRCh38
rs961717425 Chr.1: 156752135-156752135 on Build GRCh38
rs777974702 Chr.1: 156752232-156752232 on Build GRCh38
rs576764252 Chr.1: 156752294-156752294 on Build GRCh38
rs922877143 Chr.1: 156752189-156752189 on Build GRCh38
rs1038250908 Chr.1: 156752241-156752241 on Build GRCh38
rs1051304165 Chr.1: 156752228-156752228 on Build GRCh38
rs749417275 Chr.1: 156752234-156752234 on Build GRCh38
rs148762451 Chr.1: 156752201-156752201 on Build GRCh38
rs776848216 Chr.1: 156752233-156752234 on Build GRCh38
rs779903170 Chr.1: 156752147-156752147 on Build GRCh38
rs564793288 Chr.1: 156752273-156752273 on Build GRCh38
rs571979156 Chr.1: 156752139-156752139 on Build GRCh38
rs972742774 Chr.1: 156752166-156752166 on Build GRCh38
rs914821454 Chr.1: 156752197-156752197 on Build GRCh38
rs990299645 Chr.1: 156752092-156752092 on Build GRCh38
rs202106362 Chr.1: 156752223-156752223 on Build GRCh38
rs889574532 Chr.1: 156752232-156752233 on Build GRCh38
rs946877790 Chr.1: 156752237-156752237 on Build GRCh38
rs1043493739 Chr.1: 156752238-156752238 on Build GRCh38
rs1017023723 Chr.1: 156752130-156752130 on Build GRCh38
rs553808858 Chr.1: 156752093-156752093 on Build GRCh38
rs977691278 Chr.1: 156752205-156752205 on Build GRCh38
rs902241764 Chr.1: 156752310-156752310 on Build GRCh38
rs765108696 Chr.1: 156752154-156752154 on Build GRCh38
rs898408273 Chr.1: 156752314-156752316 on Build GRCh38
rs985635317 Chr.1: 156752207-156752207 on Build GRCh38
rs923609473 Chr.1: 156752221-156752221 on Build GRCh38
rs765119529 Chr.1: 156752123-156752123 on Build GRCh38
rs867680414 Chr.1: 156752085-156752085 on Build GRCh38

View Full Product Details