Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42163983-42164208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH4
Entrez Gene ID
23415
Gene Name
potassium voltage-gated channel subfamily H member 4
Gene Aliases
BEC2, ELK1, Kv12.3
UniGene
Hs.304081
Gene Chromosome Location
Chr.17: 42156892-42181278 on Build GRCh38
Transcripts
NM_012285.2 XM_017024400.1 XM_017024401.1 XM_017024402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7373041 c.2096G>A p.R699H Chr.17: 42164158-42164158 on Build GRCh38
4849306 c.2088C>T p.G696G Chr.17: 42164166-42164166 on Build GRCh38
6032982 c.2124+2T>C p.? Chr.17: 42164128-42164128 on Build GRCh38
5677721 c.2086-3C>T p.? Chr.17: 42164171-42164171 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs894114746 Chr.17: 42164007-42164007 on Build GRCh38
rs559829094 Chr.17: 42164171-42164172 on Build GRCh38
rs998648099 Chr.17: 42164030-42164030 on Build GRCh38
rs139161684 Chr.17: 42164159-42164159 on Build GRCh38
rs867480854 Chr.17: 42164149-42164149 on Build GRCh38
rs977225489 Chr.17: 42164048-42164048 on Build GRCh38
rs770057722 Chr.17: 42164158-42164158 on Build GRCh38
rs377112664 Chr.17: 42164140-42164140 on Build GRCh38
rs144714469 Chr.17: 42164116-42164116 on Build GRCh38
rs910098037 Chr.17: 42164087-42164087 on Build GRCh38
rs1021522502 Chr.17: 42164018-42164018 on Build GRCh38
rs954156170 Chr.17: 42164065-42164065 on Build GRCh38
rs563516399 Chr.17: 42164150-42164150 on Build GRCh38
rs867338987 Chr.17: 42164032-42164032 on Build GRCh38
rs201984974 Chr.17: 42164141-42164141 on Build GRCh38
rs1028345186 Chr.17: 42164063-42164063 on Build GRCh38
rs375439821 Chr.17: 42164172-42164173 on Build GRCh38
rs551094572 Chr.17: 42164090-42164090 on Build GRCh38
rs372927433 Chr.17: 42164085-42164085 on Build GRCh38
rs925174666 Chr.17: 42164170-42164170 on Build GRCh38
rs965522692 Chr.17: 42164033-42164033 on Build GRCh38
rs904363216 Chr.17: 42164025-42164025 on Build GRCh38
rs563282480 Chr.17: 42164138-42164138 on Build GRCh38
rs376807154 Chr.17: 42164182-42164182 on Build GRCh38
rs139637101 Chr.17: 42164185-42164185 on Build GRCh38
rs771082427 Chr.17: 42164181-42164181 on Build GRCh38
rs755846096 Chr.17: 42164091-42164091 on Build GRCh38
rs77835488 Chr.17: 42164013-42164013 on Build GRCh38

View Full Product Details