Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162267013-162267274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIH1
Entrez Gene ID
64135
Gene Name
interferon induced with helicase C domain 1
Gene Aliases
AGS7, Hlcd, IDDM19, MDA-5, MDA5, RLR-2, SGMRT1
UniGene
Hs.163173
Gene Chromosome Location
Chr.2: 162267079-162318708 on Build GRCh38
Transcripts
NM_022168.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1008372 c.3054C>T p.C1018C Chr.2: 162267224-162267224 on Build GRCh38
4185737 c.3040G>C p.D1014H Chr.2: 162267238-162267238 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs11891191 Chr.2: 162267194-162267194 on Build GRCh38
rs547782932 Chr.2: 162267234-162267234 on Build GRCh38
rs35369989 Chr.2: 162267236-162267236 on Build GRCh38
rs763412137 Chr.2: 162267159-162267159 on Build GRCh38
rs750808448 Chr.2: 162267157-162267157 on Build GRCh38
rs779418881 Chr.2: 162267043-162267043 on Build GRCh38
rs936730847 Chr.2: 162267065-162267065 on Build GRCh38
rs545477353 Chr.2: 162267156-162267156 on Build GRCh38
rs749016009 Chr.2: 162267149-162267149 on Build GRCh38
rs777469992 Chr.2: 162267225-162267225 on Build GRCh38
rs968646966 Chr.2: 162267058-162267058 on Build GRCh38
rs755779570 Chr.2: 162267218-162267218 on Build GRCh38
rs931567064 Chr.2: 162267243-162267243 on Build GRCh38
rs990863437 Chr.2: 162267094-162267094 on Build GRCh38
rs949389529 Chr.2: 162267179-162267179 on Build GRCh38
rs762068491 Chr.2: 162267152-162267152 on Build GRCh38
rs371793426 Chr.2: 162267191-162267191 on Build GRCh38
rs528639734 Chr.2: 162267242-162267242 on Build GRCh38
rs978550427 Chr.2: 162267047-162267047 on Build GRCh38
rs752133326 Chr.2: 162267190-162267190 on Build GRCh38
rs530560094 Chr.2: 162267108-162267108 on Build GRCh38
rs938115898 Chr.2: 162267087-162267087 on Build GRCh38
rs990932072 Chr.2: 162267130-162267130 on Build GRCh38
rs749018506 Chr.2: 162267230-162267230 on Build GRCh38
rs927102773 Chr.2: 162267085-162267085 on Build GRCh38
rs760571400 Chr.2: 162267193-162267193 on Build GRCh38
rs563135418 Chr.2: 162267054-162267054 on Build GRCh38
rs532533646 Chr.2: 162267209-162267209 on Build GRCh38
rs925813270 Chr.2: 162267050-162267050 on Build GRCh38
rs766932655 Chr.2: 162267172-162267172 on Build GRCh38
rs541763686 Chr.2: 162267037-162267037 on Build GRCh38

View Full Product Details