Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48597725-48597994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB-AS3
Entrez Gene ID
404266
Gene Name
HOXB cluster antisense RNA 3
Gene Aliases
-
UniGene
Hs.660088
Gene Chromosome Location
Chr.17: 48590420-48606414 on Build GRCh38
Transcripts
NR_033201.2 NR_033202.2 NR_033203.1 NR_033204.2 NR_033205.1 NR_110329.1 NR_110330.1 NR_110331.1 NM_018952.4 XM_005257283.3 XM_005257284.3 XM_011524727.2

Gene Symbol
HOXB6
Entrez Gene ID
3216
Gene Name
homeobox B6
Gene Aliases
HOX2, HOX2B, HU-2, Hox-2.2
UniGene
Hs.98428
Gene Chromosome Location
Chr.17: 48595736-48604992 on Build GRCh38
Transcripts
NR_033201.2 NR_033202.2 NR_033203.1 NR_033204.2 NR_033205.1 NR_110329.1 NR_110330.1 NR_110331.1 NM_018952.4 XM_005257283.3 XM_005257284.3 XM_011524727.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371430 c.331G>T p.D111Y Chr.17: 48597820-48597820 on Build GRCh38
5572799 c.198C>T p.Y66Y Chr.17: 48597953-48597953 on Build GRCh38
6328842 c.363G>T p.Q121H Chr.17: 48597788-48597788 on Build GRCh38
7291101 c.189C>G p.G63G Chr.17: 48597962-48597962 on Build GRCh38
4890214 c.288G>T p.Q96H Chr.17: 48597863-48597863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756336721 Chr.17: 48597891-48597891 on Build GRCh38
rs760358158 Chr.17: 48597869-48597869 on Build GRCh38
rs768603811 Chr.17: 48597859-48597859 on Build GRCh38
rs767983153 Chr.17: 48597799-48597799 on Build GRCh38
rs767827563 Chr.17: 48597883-48597883 on Build GRCh38
rs945286993 Chr.17: 48597923-48597923 on Build GRCh38
rs750755439 Chr.17: 48597958-48597958 on Build GRCh38
rs778154907 Chr.17: 48597749-48597749 on Build GRCh38
rs35227248 Chr.17: 48597872-48597872 on Build GRCh38
rs563745387 Chr.17: 48597901-48597901 on Build GRCh38
rs780034243 Chr.17: 48597898-48597898 on Build GRCh38
rs745936794 Chr.17: 48597783-48597783 on Build GRCh38
rs749208929 Chr.17: 48597789-48597789 on Build GRCh38
rs765144613 Chr.17: 48597841-48597843 on Build GRCh38
rs756215326 Chr.17: 48597852-48597852 on Build GRCh38
rs772912870 Chr.17: 48597812-48597812 on Build GRCh38
rs760868837 Chr.17: 48597972-48597972 on Build GRCh38
rs753170237 Chr.17: 48597822-48597822 on Build GRCh38
rs777692618 Chr.17: 48597915-48597915 on Build GRCh38
rs34438598 Chr.17: 48597878-48597878 on Build GRCh38
rs778919584 Chr.17: 48597902-48597902 on Build GRCh38
rs757440393 Chr.17: 48597776-48597776 on Build GRCh38
rs764651190 Chr.17: 48597874-48597874 on Build GRCh38
rs746021862 Chr.17: 48597850-48597850 on Build GRCh38
rs770613987 Chr.17: 48597921-48597921 on Build GRCh38
rs149102251 Chr.17: 48597813-48597813 on Build GRCh38
rs201059637 Chr.17: 48597828-48597828 on Build GRCh38
rs780133014 Chr.17: 48597854-48597854 on Build GRCh38
rs762256929 Chr.17: 48597953-48597953 on Build GRCh38
rs767272079 Chr.17: 48597811-48597811 on Build GRCh38
rs200398618 Chr.17: 48597827-48597827 on Build GRCh38
rs953731623 Chr.17: 48597936-48597936 on Build GRCh38
rs200628196 Chr.17: 48597817-48597817 on Build GRCh38
rs753944537 Chr.17: 48597900-48597900 on Build GRCh38
rs768357144 Chr.17: 48597798-48597798 on Build GRCh38
rs929297284 Chr.17: 48597773-48597773 on Build GRCh38
rs760268623 Chr.17: 48597815-48597815 on Build GRCh38
rs777522253 Chr.17: 48597917-48597917 on Build GRCh38
rs748061258 Chr.17: 48597906-48597906 on Build GRCh38
rs767917909 Chr.17: 48597955-48597955 on Build GRCh38
rs776063320 Chr.17: 48597940-48597940 on Build GRCh38
rs372308259 Chr.17: 48597809-48597809 on Build GRCh38
rs1042187599 Chr.17: 48597961-48597961 on Build GRCh38
rs546181040 Chr.17: 48597948-48597948 on Build GRCh38
rs891689463 Chr.17: 48597905-48597905 on Build GRCh38
rs746703653 Chr.17: 48597920-48597920 on Build GRCh38
rs752007409 Chr.17: 48597876-48597876 on Build GRCh38
rs747940560 Chr.17: 48597860-48597860 on Build GRCh38
rs750665681 Chr.17: 48597890-48597890 on Build GRCh38
rs771932824 Chr.17: 48597861-48597861 on Build GRCh38
rs747245099 Chr.17: 48597765-48597765 on Build GRCh38
rs769805775 Chr.17: 48597784-48597784 on Build GRCh38
rs34860593 Chr.17: 48597773-48597774 on Build GRCh38
rs370966566 Chr.17: 48597952-48597952 on Build GRCh38
rs980912425 Chr.17: 48597959-48597959 on Build GRCh38
rs577327660 Chr.17: 48597914-48597914 on Build GRCh38
rs775292723 Chr.17: 48597788-48597788 on Build GRCh38
rs773033635 Chr.17: 48597864-48597864 on Build GRCh38
rs781318489 Chr.17: 48597781-48597781 on Build GRCh38
rs534309856 Chr.17: 48597824-48597824 on Build GRCh38
rs749231927 Chr.17: 48597858-48597858 on Build GRCh38
rs143197683 Chr.17: 48597836-48597836 on Build GRCh38
rs758723592 Chr.17: 48597746-48597746 on Build GRCh38

View Full Product Details