Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62717130-62717355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL2
Entrez Gene ID
221092
Gene Name
heterogeneous nuclear ribonucleoprotein U like 2
Gene Aliases
HNRPUL2, SAF-A2
UniGene
Hs.657058
Gene Chromosome Location
Chr.11: 62712625-62727385 on Build GRCh38
Transcripts
NM_001079559.2 NR_037946.1

Gene Symbol
HNRNPUL2-BSCL2
Entrez Gene ID
100534595
Gene Name
HNRNPUL2-BSCL2 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.533709
Gene Chromosome Location
Chr.11: 62690262-62727384 on Build GRCh38
Transcripts
NM_001079559.2 NR_037946.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4886481 c.1807G>T p.D603Y Chr.11: 62717163-62717163 on Build GRCh38
1746403 c.1815G>T p.M605I Chr.11: 62717155-62717155 on Build GRCh38
6681479 c.1824G>C p.V608V Chr.11: 62717146-62717146 on Build GRCh38
929796 c.1806C>T p.C602C Chr.11: 62717164-62717164 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs937323095 Chr.11: 62717317-62717317 on Build GRCh38
rs752374816 Chr.11: 62717196-62717196 on Build GRCh38
rs753580483 Chr.11: 62717188-62717188 on Build GRCh38
rs370181150 Chr.11: 62717237-62717237 on Build GRCh38
rs530320304 Chr.11: 62717223-62717223 on Build GRCh38
rs950329350 Chr.11: 62717264-62717264 on Build GRCh38
rs772743752 Chr.11: 62717159-62717159 on Build GRCh38
rs1029725859 Chr.11: 62717151-62717151 on Build GRCh38
rs373909806 Chr.11: 62717157-62717157 on Build GRCh38
rs758381191 Chr.11: 62717198-62717198 on Build GRCh38
rs993750793 Chr.11: 62717248-62717248 on Build GRCh38
rs925673165 Chr.11: 62717212-62717212 on Build GRCh38
rs913430292 Chr.11: 62717238-62717238 on Build GRCh38
rs376859902 Chr.11: 62717193-62717193 on Build GRCh38
rs567158531 Chr.11: 62717267-62717267 on Build GRCh38
rs113109055 Chr.11: 62717192-62717192 on Build GRCh38
rs771131249 Chr.11: 62717324-62717324 on Build GRCh38
rs765857560 Chr.11: 62717179-62717179 on Build GRCh38
rs1049094260 Chr.11: 62717325-62717325 on Build GRCh38
rs760222315 Chr.11: 62717164-62717164 on Build GRCh38
rs1040964450 Chr.11: 62717160-62717160 on Build GRCh38
rs769104355 Chr.11: 62717150-62717150 on Build GRCh38
rs192018309 Chr.11: 62717233-62717233 on Build GRCh38
rs1022733654 Chr.11: 62717316-62717316 on Build GRCh38
rs550320915 Chr.11: 62717231-62717231 on Build GRCh38
rs552336862 Chr.11: 62717300-62717300 on Build GRCh38
rs776098855 Chr.11: 62717264-62717266 on Build GRCh38
rs751510005 Chr.11: 62717220-62717220 on Build GRCh38
rs560922145 Chr.11: 62717206-62717206 on Build GRCh38
rs985608786 Chr.11: 62717180-62717180 on Build GRCh38

View Full Product Details