Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138559699-138559924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSPA9
Entrez Gene ID
3313
Gene Name
heat shock protein family A (Hsp70) member 9
Gene Aliases
CRP40, CSA, EVPLS, GRP-75, GRP75, HEL-S-124m, HSPA9B, MOT, MOT2, MTHSP75, PBP74, SAAN, SIDBA4
UniGene
Hs.184233
Gene Chromosome Location
Chr.5: 138554882-138575629 on Build GRCh38
Transcripts
NM_004134.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4625411 c.1407C>T p.S469S Chr.5: 138559867-138559867 on Build GRCh38
1619533 c.1378A>C p.R460R Chr.5: 138559896-138559896 on Build GRCh38
7313071 c.1350A>G p.L450L Chr.5: 138559924-138559924 on Build GRCh38
7334138 c.1410+6_1410+7delAG p.? Chr.5: 138559857-138559858 on Build GRCh38
3850466 c.1387A>G p.T463A Chr.5: 138559887-138559887 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779394970 Chr.5: 138559818-138559818 on Build GRCh38
rs775053732 Chr.5: 138559807-138559807 on Build GRCh38
rs1000522964 Chr.5: 138559805-138559805 on Build GRCh38
rs538667398 Chr.5: 138559801-138559801 on Build GRCh38
rs749918069 Chr.5: 138559813-138559813 on Build GRCh38
rs778181446 Chr.5: 138559836-138559836 on Build GRCh38
rs369690198 Chr.5: 138559876-138559876 on Build GRCh38
rs553787114 Chr.5: 138559808-138559808 on Build GRCh38
rs972968585 Chr.5: 138559887-138559887 on Build GRCh38
rs781187826 Chr.5: 138559873-138559873 on Build GRCh38
rs142975569 Chr.5: 138559778-138559778 on Build GRCh38
rs369901193 Chr.5: 138559814-138559814 on Build GRCh38
rs747239177 Chr.5: 138559844-138559844 on Build GRCh38
rs374257815 Chr.5: 138559835-138559835 on Build GRCh38
rs144313015 Chr.5: 138559849-138559849 on Build GRCh38
rs555443921 Chr.5: 138559759-138559760 on Build GRCh38
rs902145173 Chr.5: 138559792-138559792 on Build GRCh38
rs567030218 Chr.5: 138559767-138559767 on Build GRCh38
rs753769082 Chr.5: 138559831-138559831 on Build GRCh38
rs1019749684 Chr.5: 138559868-138559868 on Build GRCh38

View Full Product Details