Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22605651-22605924 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCAR2
Entrez Gene ID
57805
Gene Name
cell cycle and apoptosis regulator 2
Gene Aliases
DBC-1, DBC1, KIAA1967, NET35, p30 DBC, p30DBC
UniGene
Hs.744848
Gene Chromosome Location
Chr.8: 22604632-22620471 on Build GRCh38
Transcripts
NM_021174.5 NR_033902.1 XM_011544603.1 XM_011544604.1 XM_017013717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770937 c.58+3G>A p.? Chr.8: 22605834-22605834 on Build GRCh38
229753 c.21G>C p.Q7H Chr.8: 22605794-22605794 on Build GRCh38
1098128 c.4T>C p.S2P Chr.8: 22605777-22605777 on Build GRCh38
6694498 c.17G>A p.R6H Chr.8: 22605790-22605790 on Build GRCh38
3699024 c.47G>A p.R16H Chr.8: 22605820-22605820 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755808796 Chr.8: 22605770-22605770 on Build GRCh38
rs200669776 Chr.8: 22605840-22605840 on Build GRCh38
rs570271006 Chr.8: 22605901-22605901 on Build GRCh38
rs371733204 Chr.8: 22605790-22605790 on Build GRCh38
rs150400388 Chr.8: 22605806-22605806 on Build GRCh38
rs761301974 Chr.8: 22605836-22605836 on Build GRCh38
rs778465706 Chr.8: 22605883-22605886 on Build GRCh38
rs200324788 Chr.8: 22605721-22605721 on Build GRCh38
rs377252239 Chr.8: 22605729-22605729 on Build GRCh38
rs201084892 Chr.8: 22605733-22605733 on Build GRCh38
rs201039859 Chr.8: 22605873-22605873 on Build GRCh38
rs775799558 Chr.8: 22605822-22605822 on Build GRCh38
rs745901830 Chr.8: 22605804-22605804 on Build GRCh38
rs751121974 Chr.8: 22605743-22605743 on Build GRCh38
rs780956169 Chr.8: 22605754-22605754 on Build GRCh38
rs756479105 Chr.8: 22605706-22605706 on Build GRCh38
rs984725411 Chr.8: 22605682-22605682 on Build GRCh38
rs199498908 Chr.8: 22605781-22605781 on Build GRCh38
rs199877708 Chr.8: 22605712-22605712 on Build GRCh38
rs373808946 Chr.8: 22605844-22605844 on Build GRCh38
rs777683635 Chr.8: 22605779-22605779 on Build GRCh38
rs754510741 Chr.8: 22605746-22605746 on Build GRCh38
rs767072187 Chr.8: 22605879-22605879 on Build GRCh38
rs762658295 Chr.8: 22605737-22605737 on Build GRCh38
rs201769711 Chr.8: 22605802-22605802 on Build GRCh38
rs558909685 Chr.8: 22605758-22605758 on Build GRCh38
rs910945472 Chr.8: 22605899-22605899 on Build GRCh38
rs1037594609 Chr.8: 22605717-22605717 on Build GRCh38
rs200104791 Chr.8: 22605872-22605872 on Build GRCh38
rs1051319120 Chr.8: 22605756-22605756 on Build GRCh38
rs762574353 Chr.8: 22605852-22605852 on Build GRCh38
rs756323589 Chr.8: 22605819-22605819 on Build GRCh38
rs765847314 Chr.8: 22605742-22605742 on Build GRCh38
rs200959219 Chr.8: 22605795-22605795 on Build GRCh38
rs765968605 Chr.8: 22605869-22605869 on Build GRCh38
rs752030174 Chr.8: 22605882-22605882 on Build GRCh38
rs201673414 Chr.8: 22605850-22605850 on Build GRCh38
rs200406449 Chr.8: 22605702-22605702 on Build GRCh38
rs74608631 Chr.8: 22605683-22605683 on Build GRCh38
rs764528711 Chr.8: 22605728-22605728 on Build GRCh38
rs201441237 Chr.8: 22605703-22605703 on Build GRCh38
rs201715739 Chr.8: 22605876-22605876 on Build GRCh38
rs369800538 Chr.8: 22605744-22605744 on Build GRCh38

View Full Product Details