Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63624115-63624370 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC1
Entrez Gene ID
8925
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Gene Aliases
p532, p619
UniGene
Hs.210385
Gene Chromosome Location
Chr.15: 63608618-63833964 on Build GRCh38
Transcripts
NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4819873 c.13276-1G>C p.? Chr.15: 63624328-63624328 on Build GRCh38
5483939 c.13310G>A p.G4437D Chr.15: 63624293-63624293 on Build GRCh38
3794304 c.13334C>A p.P4445H Chr.15: 63624269-63624269 on Build GRCh38
2255021 c.13322G>A p.G4441E Chr.15: 63624281-63624281 on Build GRCh38
5980776 c.13312A>G p.I4438V Chr.15: 63624291-63624291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781139859 Chr.15: 63624331-63624331 on Build GRCh38
rs756289514 Chr.15: 63624265-63624265 on Build GRCh38
rs773560667 Chr.15: 63624237-63624237 on Build GRCh38
rs377137482 Chr.15: 63624277-63624277 on Build GRCh38
rs753722310 Chr.15: 63624269-63624269 on Build GRCh38
rs368224538 Chr.15: 63624214-63624214 on Build GRCh38
rs763224069 Chr.15: 63624246-63624246 on Build GRCh38
rs1004508684 Chr.15: 63624162-63624162 on Build GRCh38
rs536068959 Chr.15: 63624310-63624310 on Build GRCh38
rs953020446 Chr.15: 63624294-63624294 on Build GRCh38
rs767041049 Chr.15: 63624247-63624247 on Build GRCh38
rs770067924 Chr.15: 63624231-63624231 on Build GRCh38
rs775039073 Chr.15: 63624225-63624225 on Build GRCh38
rs372316431 Chr.15: 63624230-63624230 on Build GRCh38
rs778812222 Chr.15: 63624187-63624187 on Build GRCh38
rs770917843 Chr.15: 63624173-63624173 on Build GRCh38
rs201872684 Chr.15: 63624272-63624272 on Build GRCh38
rs764214583 Chr.15: 63624266-63624266 on Build GRCh38
rs749708623 Chr.15: 63624343-63624343 on Build GRCh38
rs776124619 Chr.15: 63624336-63624336 on Build GRCh38
rs757927407 Chr.15: 63624291-63624291 on Build GRCh38
rs752871809 Chr.15: 63624261-63624261 on Build GRCh38
rs371859723 Chr.15: 63624168-63624168 on Build GRCh38
rs375298173 Chr.15: 63624199-63624199 on Build GRCh38
rs554429475 Chr.15: 63624330-63624330 on Build GRCh38
rs749182762 Chr.15: 63624151-63624151 on Build GRCh38
rs200325012 Chr.15: 63624308-63624308 on Build GRCh38
rs753617184 Chr.15: 63624190-63624190 on Build GRCh38
rs759720013 Chr.15: 63624259-63624259 on Build GRCh38
rs997637254 Chr.15: 63624245-63624245 on Build GRCh38
rs777563355 Chr.15: 63624144-63624144 on Build GRCh38
rs907472110 Chr.15: 63624143-63624143 on Build GRCh38
rs752080004 Chr.15: 63624255-63624255 on Build GRCh38

View Full Product Details