Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140654483-140654710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IK
Entrez Gene ID
3550
Gene Name
IK cytokine, down-regulator of HLA II
Gene Aliases
CSA2, RED, RER
UniGene
Hs.421245
Gene Chromosome Location
Chr.5: 140647799-140662480 on Build GRCh38
Transcripts
NM_006083.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3726462 c.566T>C p.M189T Chr.5: 140654562-140654562 on Build GRCh38
4450281 c.520-1G>T p.? Chr.5: 140654515-140654515 on Build GRCh38
1061735 c.610A>C p.N204H Chr.5: 140654700-140654700 on Build GRCh38
3994011 c.555A>C p.E185D Chr.5: 140654551-140654551 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754208095 Chr.5: 140654645-140654645 on Build GRCh38
rs868619559 Chr.5: 140654540-140654540 on Build GRCh38
rs1047849625 Chr.5: 140654646-140654646 on Build GRCh38
rs767703715 Chr.5: 140654584-140654584 on Build GRCh38
rs138578964 Chr.5: 140654617-140654617 on Build GRCh38
rs373920549 Chr.5: 140654649-140654649 on Build GRCh38
rs969436844 Chr.5: 140654548-140654548 on Build GRCh38
rs200201511 Chr.5: 140654534-140654534 on Build GRCh38
rs752544653 Chr.5: 140654600-140654600 on Build GRCh38
rs755749141 Chr.5: 140654667-140654667 on Build GRCh38
rs767538550 Chr.5: 140654545-140654545 on Build GRCh38
rs970366958 Chr.5: 140654504-140654504 on Build GRCh38
rs747687273 Chr.5: 140654653-140654654 on Build GRCh38
rs756037619 Chr.5: 140654603-140654603 on Build GRCh38
rs774318672 Chr.5: 140654547-140654547 on Build GRCh38
rs771065050 Chr.5: 140654638-140654649 on Build GRCh38
rs760741330 Chr.5: 140654616-140654617 on Build GRCh38
rs754436450 Chr.5: 140654653-140654653 on Build GRCh38
rs930591953 Chr.5: 140654634-140654634 on Build GRCh38
rs944895780 Chr.5: 140654672-140654672 on Build GRCh38
rs775560681 Chr.5: 140654605-140654605 on Build GRCh38
rs749109847 Chr.5: 140654535-140654535 on Build GRCh38
rs764224644 Chr.5: 140654625-140654625 on Build GRCh38
rs1003175656 Chr.5: 140654568-140654568 on Build GRCh38
rs769348315 Chr.5: 140654530-140654530 on Build GRCh38
rs762319017 Chr.5: 140654647-140654647 on Build GRCh38
rs201887185 Chr.5: 140654519-140654519 on Build GRCh38
rs759576691 Chr.5: 140654554-140654554 on Build GRCh38
rs561634572 Chr.5: 140654665-140654665 on Build GRCh38
rs760892324 Chr.5: 140654611-140654611 on Build GRCh38
rs772995921 Chr.5: 140654531-140654531 on Build GRCh38

View Full Product Details