Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123655770-123656029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2H3
Entrez Gene ID
2967
Gene Name
general transcription factor IIH subunit 3
Gene Aliases
BTF2, P34, TFB4, TFIIH
UniGene
Hs.355348
Gene Chromosome Location
Chr.12: 123633739-123662604 on Build GRCh38
Transcripts
NM_001271866.1 NM_001271867.1 NM_001271868.1 NM_001516.4 XM_017019228.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6437256 c.587T>C p.L196S Chr.12: 123655796-123655796 on Build GRCh38
6676712 c.568T>G p.L190V Chr.12: 123655777-123655777 on Build GRCh38
6332783 c.615+5G>T p.? Chr.12: 123655829-123655829 on Build GRCh38
1605721 c.583G>T p.V195F Chr.12: 123655792-123655792 on Build GRCh38
6355902 c.564T>C p.N188N Chr.12: 123655773-123655773 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976952172 Chr.12: 123655921-123655921 on Build GRCh38
rs770019312 Chr.12: 123655808-123655808 on Build GRCh38
rs774994175 Chr.12: 123655818-123655818 on Build GRCh38
rs746112121 Chr.12: 123655804-123655804 on Build GRCh38
rs954313554 Chr.12: 123655932-123655932 on Build GRCh38
rs11057318 Chr.12: 123655941-123655941 on Build GRCh38
rs776888139 Chr.12: 123655803-123655803 on Build GRCh38
rs184841046 Chr.12: 123655884-123655884 on Build GRCh38
rs1054974121 Chr.12: 123656002-123656002 on Build GRCh38
rs1011119373 Chr.12: 123655802-123655802 on Build GRCh38
rs371746742 Chr.12: 123655852-123655852 on Build GRCh38
rs1052350294 Chr.12: 123655972-123655972 on Build GRCh38
rs11057319 Chr.12: 123655965-123655965 on Build GRCh38
rs1031775653 Chr.12: 123655898-123655898 on Build GRCh38
rs777731993 Chr.12: 123655869-123655869 on Build GRCh38
rs765961953 Chr.12: 123655851-123655851 on Build GRCh38
rs11572986 Chr.12: 123655988-123655988 on Build GRCh38
rs918292130 Chr.12: 123655912-123655912 on Build GRCh38
rs1039561697 Chr.12: 123655955-123655955 on Build GRCh38
rs972426358 Chr.12: 123655971-123655971 on Build GRCh38
rs763316736 Chr.12: 123655811-123655811 on Build GRCh38
rs772714207 Chr.12: 123655936-123655936 on Build GRCh38
rs764565875 Chr.12: 123655813-123655813 on Build GRCh38
rs909534429 Chr.12: 123655953-123655953 on Build GRCh38
rs199891907 Chr.12: 123655810-123655810 on Build GRCh38
rs555600805 Chr.12: 123655931-123655931 on Build GRCh38
rs375137857 Chr.12: 123655840-123655840 on Build GRCh38
rs11572984 Chr.12: 123655855-123655855 on Build GRCh38
rs959802827 Chr.12: 123655865-123655865 on Build GRCh38
rs11572985 Chr.12: 123655856-123655856 on Build GRCh38
rs901262697 Chr.12: 123655963-123655963 on Build GRCh38
rs189770703 Chr.12: 123655918-123655918 on Build GRCh38
rs942513035 Chr.12: 123655943-123655943 on Build GRCh38
rs758212913 Chr.12: 123655860-123655860 on Build GRCh38

View Full Product Details