Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46312120-46312389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIF3A
Entrez Gene ID
64344
Gene Name
hypoxia inducible factor 3 alpha subunit
Gene Aliases
HIF-3A, HIF3-alpha-1, IPAS, MOP7, PASD7, bHLHe17
UniGene
Hs.420830
Gene Chromosome Location
Chr.19: 46297046-46343433 on Build GRCh38
Transcripts
NM_022462.4 NM_152794.3 NM_152795.3 NM_152796.2 XM_005259152.4 XM_005259153.3 XM_005259155.4 XM_005259156.4 XM_017027132.1 XM_017027133.1 XM_017027134.1 XM_017027135.1 XM_017027136.1 XM_017027137.1 XM_017027138.1 XM_017027139.1 XM_017027140.1 XM_017027141.1 XM_017027142.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311772 c.782G>A p.G261D Chr.19: 46312178-46312178 on Build GRCh38
5707086 c.685G>A p.A229T Chr.19: 46312228-46312228 on Build GRCh38
5635769 c.845A>C p.D282A Chr.19: 46312241-46312241 on Build GRCh38
3371383 c.781G>T p.V261L Chr.19: 46312171-46312171 on Build GRCh38
5659551 c.635G>A p.G212D Chr.19: 46312178-46312178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763822130 Chr.19: 46312173-46312173 on Build GRCh38
rs528381669 Chr.19: 46312327-46312327 on Build GRCh38
rs768560350 Chr.19: 46312307-46312307 on Build GRCh38
rs987345030 Chr.19: 46312278-46312278 on Build GRCh38
rs746992488 Chr.19: 46312298-46312298 on Build GRCh38
rs772020306 Chr.19: 46312152-46312152 on Build GRCh38
rs769512846 Chr.19: 46312315-46312315 on Build GRCh38
rs768157916 Chr.19: 46312343-46312343 on Build GRCh38
rs776380721 Chr.19: 46312230-46312230 on Build GRCh38
rs1010742168 Chr.19: 46312251-46312251 on Build GRCh38
rs773701309 Chr.19: 46312334-46312334 on Build GRCh38
rs565680427 Chr.19: 46312313-46312313 on Build GRCh38
rs766941547 Chr.19: 46312192-46312192 on Build GRCh38
rs770540295 Chr.19: 46312332-46312332 on Build GRCh38
rs944166463 Chr.19: 46312143-46312143 on Build GRCh38
rs145550030 Chr.19: 46312267-46312267 on Build GRCh38
rs781679049 Chr.19: 46312201-46312201 on Build GRCh38
rs1049018793 Chr.19: 46312219-46312219 on Build GRCh38
rs748320920 Chr.19: 46312141-46312141 on Build GRCh38
rs181266954 Chr.19: 46312292-46312292 on Build GRCh38
rs8101480 Chr.19: 46312242-46312242 on Build GRCh38
rs559320651 Chr.19: 46312147-46312147 on Build GRCh38
rs200909095 Chr.19: 46312229-46312229 on Build GRCh38
rs761001559 Chr.19: 46312145-46312146 on Build GRCh38
rs201426103 Chr.19: 46312200-46312200 on Build GRCh38
rs374324091 Chr.19: 46312217-46312217 on Build GRCh38
rs1016723764 Chr.19: 46312146-46312146 on Build GRCh38
rs764768309 Chr.19: 46312189-46312189 on Build GRCh38
rs751943658 Chr.19: 46312195-46312195 on Build GRCh38
rs760527371 Chr.19: 46312170-46312170 on Build GRCh38
rs753953307 Chr.19: 46312274-46312274 on Build GRCh38
rs778083512 Chr.19: 46312271-46312271 on Build GRCh38
rs762320739 Chr.19: 46312191-46312191 on Build GRCh38
rs778045317 Chr.19: 46312216-46312216 on Build GRCh38
rs762528091 Chr.19: 46312244-46312244 on Build GRCh38
rs75205676 Chr.19: 46312227-46312227 on Build GRCh38
rs543522717 Chr.19: 46312239-46312239 on Build GRCh38
rs764357342 Chr.19: 46312366-46312366 on Build GRCh38
rs375306480 Chr.19: 46312308-46312308 on Build GRCh38
rs760114983 Chr.19: 46312246-46312246 on Build GRCh38
rs548033317 Chr.19: 46312354-46312354 on Build GRCh38
rs374922972 Chr.19: 46312154-46312154 on Build GRCh38
rs769459593 Chr.19: 46312240-46312240 on Build GRCh38
rs572671763 Chr.19: 46312148-46312148 on Build GRCh38
rs753034506 Chr.19: 46312210-46312210 on Build GRCh38
rs778857461 Chr.19: 46312293-46312293 on Build GRCh38
rs761123054 Chr.19: 46312361-46312361 on Build GRCh38
rs776096601 Chr.19: 46312350-46312350 on Build GRCh38
rs371315415 Chr.19: 46312188-46312188 on Build GRCh38
rs780058400 Chr.19: 46312228-46312228 on Build GRCh38
rs147931736 Chr.19: 46312245-46312245 on Build GRCh38
rs756370923 Chr.19: 46312215-46312215 on Build GRCh38
rs753087580 Chr.19: 46312256-46312256 on Build GRCh38
rs776325232 Chr.19: 46312186-46312186 on Build GRCh38
rs768030968 Chr.19: 46312255-46312255 on Build GRCh38
rs775238932 Chr.19: 46312155-46312155 on Build GRCh38
rs749404102 Chr.19: 46312225-46312225 on Build GRCh38
rs760166352 Chr.19: 46312338-46312338 on Build GRCh38
rs762500270 Chr.19: 46312331-46312331 on Build GRCh38

View Full Product Details