Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33287023-33287260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK3
Entrez Gene ID
10114
Gene Name
homeodomain interacting protein kinase 3
Gene Aliases
DYRK6, FIST3, PKY, YAK1
UniGene
Hs.201918
Gene Chromosome Location
Chr.11: 33256672-33357023 on Build GRCh38
Transcripts
NM_001048200.2 NM_001278162.1 NM_001278163.1 NM_005734.4 XM_005252729.2 XM_017017076.1 XM_017017077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4616151 c.836A>C p.E279A Chr.11: 33287250-33287250 on Build GRCh38
3446851 c.659A>G p.N220S Chr.11: 33287073-33287073 on Build GRCh38
5970149 c.702T>C p.Y234Y Chr.11: 33287116-33287116 on Build GRCh38
6300505 c.834delG p.E279fs*8 Chr.11: 33287248-33287248 on Build GRCh38
2114981 c.721G>A p.E241K Chr.11: 33287135-33287135 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775536981 Chr.11: 33287200-33287200 on Build GRCh38
rs976152004 Chr.11: 33287127-33287127 on Build GRCh38
rs777509665 Chr.11: 33287181-33287181 on Build GRCh38
rs749256013 Chr.11: 33287182-33287182 on Build GRCh38
rs376935843 Chr.11: 33287068-33287068 on Build GRCh38
rs781029181 Chr.11: 33287138-33287138 on Build GRCh38
rs751170625 Chr.11: 33287121-33287121 on Build GRCh38
rs376939727 Chr.11: 33287216-33287216 on Build GRCh38
rs755955880 Chr.11: 33287158-33287158 on Build GRCh38
rs758924721 Chr.11: 33287114-33287115 on Build GRCh38
rs765907964 Chr.11: 33287120-33287120 on Build GRCh38
rs922016811 Chr.11: 33287139-33287139 on Build GRCh38
rs764361772 Chr.11: 33287078-33287078 on Build GRCh38
rs770844579 Chr.11: 33287185-33287185 on Build GRCh38
rs867823585 Chr.11: 33287118-33287118 on Build GRCh38
rs535328561 Chr.11: 33287098-33287098 on Build GRCh38
rs931991377 Chr.11: 33287146-33287146 on Build GRCh38
rs760884860 Chr.11: 33287073-33287073 on Build GRCh38
rs750962772 Chr.11: 33287108-33287108 on Build GRCh38
rs200282854 Chr.11: 33287226-33287226 on Build GRCh38
rs765622186 Chr.11: 33287083-33287083 on Build GRCh38
rs369914142 Chr.11: 33287048-33287048 on Build GRCh38
rs754653543 Chr.11: 33287131-33287131 on Build GRCh38
rs928188226 Chr.11: 33287198-33287198 on Build GRCh38
rs150819399 Chr.11: 33287155-33287155 on Build GRCh38
rs757779297 Chr.11: 33287111-33287111 on Build GRCh38
rs374098516 Chr.11: 33287080-33287080 on Build GRCh38
rs573350879 Chr.11: 33287163-33287163 on Build GRCh38
rs969951797 Chr.11: 33287179-33287179 on Build GRCh38
rs139279668 Chr.11: 33287217-33287217 on Build GRCh38
rs55650492 Chr.11: 33287047-33287047 on Build GRCh38
rs546084528 Chr.11: 33287149-33287149 on Build GRCh38

View Full Product Details