Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139177004-139177235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
UniGene
Hs.197644
Gene Chromosome Location
Chr.6: 139135112-139180809 on Build GRCh38
Transcripts
NM_016217.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6365021 c.1624G>A p.A542T Chr.6: 139177097-139177097 on Build GRCh38
5585408 c.1608C>T p.S536S Chr.6: 139177081-139177081 on Build GRCh38
4469646 c.1611C>T p.F537F Chr.6: 139177084-139177084 on Build GRCh38
1620983 c.1591G>T p.V531L Chr.6: 139177064-139177064 on Build GRCh38
3621137 c.1620C>A p.L540L Chr.6: 139177093-139177093 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032037086 Chr.6: 139177158-139177158 on Build GRCh38
rs998516503 Chr.6: 139177137-139177137 on Build GRCh38
rs1052283962 Chr.6: 139177085-139177085 on Build GRCh38
rs749826101 Chr.6: 139177091-139177091 on Build GRCh38
rs752905042 Chr.6: 139177101-139177101 on Build GRCh38
rs765393458 Chr.6: 139177148-139177148 on Build GRCh38
rs557285427 Chr.6: 139177081-139177081 on Build GRCh38
rs747854103 Chr.6: 139177064-139177064 on Build GRCh38
rs768947958 Chr.6: 139177130-139177130 on Build GRCh38
rs370400624 Chr.6: 139177102-139177102 on Build GRCh38
rs754794268 Chr.6: 139177057-139177057 on Build GRCh38
rs747534366 Chr.6: 139177116-139177116 on Build GRCh38
rs777457385 Chr.6: 139177086-139177086 on Build GRCh38
rs897821413 Chr.6: 139177049-139177049 on Build GRCh38
rs1007820450 Chr.6: 139177185-139177185 on Build GRCh38
rs751371934 Chr.6: 139177036-139177036 on Build GRCh38
rs775797630 Chr.6: 139177111-139177111 on Build GRCh38
rs762049934 Chr.6: 139177140-139177140 on Build GRCh38
rs201854664 Chr.6: 139177134-139177134 on Build GRCh38
rs748702173 Chr.6: 139177094-139177094 on Build GRCh38
rs767636851 Chr.6: 139177063-139177063 on Build GRCh38

View Full Product Details