Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9893267-9893508 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAGN1
Entrez Gene ID
84522
Gene Name
jagunal homolog 1
Gene Aliases
GL009, SCN6
UniGene
Hs.24054
Gene Chromosome Location
Chr.3: 9890587-9894349 on Build GRCh38
Transcripts
NM_032492.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1328028 c.513C>A p.D171E Chr.3: 9893338-9893338 on Build GRCh38
6588935 c.459G>A p.V153V Chr.3: 9893284-9893284 on Build GRCh38
1049298 c.454T>C p.L152L Chr.3: 9893279-9893279 on Build GRCh38
1211217 c.508C>A p.L170I Chr.3: 9893333-9893333 on Build GRCh38
4918479 c.490T>C p.Y164H Chr.3: 9893315-9893315 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs565971808 Chr.3: 9893433-9893433 on Build GRCh38
rs777461211 Chr.3: 9893427-9893427 on Build GRCh38
rs779442557 Chr.3: 9893305-9893305 on Build GRCh38
rs544601632 Chr.3: 9893468-9893468 on Build GRCh38
rs577126931 Chr.3: 9893454-9893454 on Build GRCh38
rs587777730 Chr.3: 9893310-9893310 on Build GRCh38
rs773059565 Chr.3: 9893324-9893324 on Build GRCh38
rs774366333 Chr.3: 9893349-9893349 on Build GRCh38
rs752200138 Chr.3: 9893413-9893413 on Build GRCh38
rs778830325 Chr.3: 9893384-9893385 on Build GRCh38
rs749140125 Chr.3: 9893356-9893356 on Build GRCh38
rs999050315 Chr.3: 9893368-9893368 on Build GRCh38
rs764403550 Chr.3: 9893358-9893358 on Build GRCh38
rs79555969 Chr.3: 9893410-9893410 on Build GRCh38
rs750948225 Chr.3: 9893408-9893408 on Build GRCh38
rs11554811 Chr.3: 9893386-9893386 on Build GRCh38
rs754191323 Chr.3: 9893366-9893366 on Build GRCh38
rs527968298 Chr.3: 9893301-9893301 on Build GRCh38
rs775653139 Chr.3: 9893357-9893357 on Build GRCh38
rs754551666 Chr.3: 9893311-9893311 on Build GRCh38
rs1043365547 Chr.3: 9893344-9893344 on Build GRCh38
rs757768981 Chr.3: 9893302-9893302 on Build GRCh38
rs201942958 Chr.3: 9893411-9893411 on Build GRCh38
rs759350997 Chr.3: 9893351-9893351 on Build GRCh38
rs61738795 Chr.3: 9893323-9893323 on Build GRCh38
rs558928728 Chr.3: 9893449-9893449 on Build GRCh38
rs370035886 Chr.3: 9893347-9893347 on Build GRCh38
rs11554812 Chr.3: 9893296-9893296 on Build GRCh38
rs944137140 Chr.3: 9893480-9893480 on Build GRCh38
rs577908383 Chr.3: 9893421-9893421 on Build GRCh38
rs919199588 Chr.3: 9893447-9893447 on Build GRCh38
rs73811601 Chr.3: 9893434-9893434 on Build GRCh38
rs538982623 Chr.3: 9893456-9893456 on Build GRCh38
rs372024121 Chr.3: 9893486-9893486 on Build GRCh38
rs372426169 Chr.3: 9893321-9893321 on Build GRCh38
rs1000072298 Chr.3: 9893314-9893314 on Build GRCh38
rs756989255 Chr.3: 9893361-9893363 on Build GRCh38
rs368135532 Chr.3: 9893441-9893441 on Build GRCh38
rs749080431 Chr.3: 9893328-9893328 on Build GRCh38
rs767709484 Chr.3: 9893356-9893356 on Build GRCh38
rs201464438 Chr.3: 9893416-9893416 on Build GRCh38

View Full Product Details