Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 138405064-138405325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEBP2
Entrez Gene ID
23593
Gene Name
heme binding protein 2
Gene Aliases
C6ORF34B, C6orf34, PP23, SOUL
UniGene
Hs.486589
Gene Chromosome Location
Chr.6: 138404199-138415593 on Build GRCh38
Transcripts
NM_001326380.1 NM_001326381.1 NM_014320.2 XM_017010631.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201124 c.238G>A p.E80K Chr.6: 138405280-138405280 on Build GRCh38
1179947 c.125A>G p.H42R Chr.6: 138405167-138405167 on Build GRCh38
6678661 c.196T>C p.F66L Chr.6: 138405238-138405238 on Build GRCh38
1195763 c.168G>A p.M56I Chr.6: 138405210-138405210 on Build GRCh38
5628145 c.132A>G p.G44G Chr.6: 138405174-138405174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053739420 Chr.6: 138405097-138405097 on Build GRCh38
rs779300426 Chr.6: 138405267-138405267 on Build GRCh38
rs746543703 Chr.6: 138405161-138405161 on Build GRCh38
rs754550383 Chr.6: 138405239-138405239 on Build GRCh38
rs1053846122 Chr.6: 138405103-138405103 on Build GRCh38
rs148433984 Chr.6: 138405146-138405146 on Build GRCh38
rs575066766 Chr.6: 138405228-138405228 on Build GRCh38
rs371279327 Chr.6: 138405263-138405263 on Build GRCh38
rs763424871 Chr.6: 138405110-138405110 on Build GRCh38
rs1041795737 Chr.6: 138405108-138405108 on Build GRCh38
rs892276206 Chr.6: 138405102-138405102 on Build GRCh38
rs752830487 Chr.6: 138405122-138405122 on Build GRCh38
rs147719512 Chr.6: 138405257-138405257 on Build GRCh38
rs75932827 Chr.6: 138405287-138405287 on Build GRCh38
rs575078159 Chr.6: 138405211-138405211 on Build GRCh38
rs765013890 Chr.6: 138405215-138405215 on Build GRCh38
rs1026658769 Chr.6: 138405220-138405220 on Build GRCh38
rs761666443 Chr.6: 138405213-138405213 on Build GRCh38
rs749419933 Chr.6: 138405144-138405144 on Build GRCh38
rs750794680 Chr.6: 138405148-138405148 on Build GRCh38
rs982465795 Chr.6: 138405258-138405258 on Build GRCh38
rs781482038 Chr.6: 138405125-138405125 on Build GRCh38
rs772406101 Chr.6: 138405279-138405279 on Build GRCh38
rs775665563 Chr.6: 138405194-138405194 on Build GRCh38
rs757995098 Chr.6: 138405226-138405226 on Build GRCh38
rs773809999 Chr.6: 138405174-138405174 on Build GRCh38
rs753542102 Chr.6: 138405138-138405138 on Build GRCh38
rs142901131 Chr.6: 138405181-138405181 on Build GRCh38
rs774593445 Chr.6: 138405186-138405186 on Build GRCh38
rs748587277 Chr.6: 138405094-138405094 on Build GRCh38
rs184504326 Chr.6: 138405105-138405105 on Build GRCh38
rs143522831 Chr.6: 138405116-138405116 on Build GRCh38
rs193920889 Chr.6: 138405167-138405167 on Build GRCh38
rs200987613 Chr.6: 138405196-138405196 on Build GRCh38
rs774703179 Chr.6: 138405119-138405119 on Build GRCh38
rs534273390 Chr.6: 138405234-138405234 on Build GRCh38
rs778131720 Chr.6: 138405248-138405248 on Build GRCh38
rs377718366 Chr.6: 138405217-138405217 on Build GRCh38
rs756926060 Chr.6: 138405124-138405125 on Build GRCh38
rs770314802 Chr.6: 138405100-138405100 on Build GRCh38
rs758049951 Chr.6: 138405160-138405160 on Build GRCh38
rs759522784 Chr.6: 138405121-138405121 on Build GRCh38
rs757137760 Chr.6: 138405139-138405139 on Build GRCh38
rs759740310 Chr.6: 138405191-138405191 on Build GRCh38
rs760890007 Chr.6: 138405195-138405195 on Build GRCh38
rs770263401 Chr.6: 138405170-138405170 on Build GRCh38
rs9495077 Chr.6: 138405131-138405131 on Build GRCh38
rs746260581 Chr.6: 138405275-138405275 on Build GRCh38

View Full Product Details