Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 13298718-13298981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISM1
Entrez Gene ID
140862
Gene Name
isthmin 1, angiogenesis inhibitor
Gene Aliases
C20orf82, ISM, Isthmin, bA149I18.1, dJ1077I2.1
UniGene
Hs.559353
Gene Chromosome Location
Chr.20: 13221771-13326734 on Build GRCh38
Transcripts
NM_080826.1 XM_017027680.1

Gene Symbol
TASP1
Entrez Gene ID
55617
Gene Name
taspase 1
Gene Aliases
C20orf13, dJ585I14.2
UniGene
Hs.348297
Gene Chromosome Location
Chr.20: 13104760-13638936 on Build GRCh38
Transcripts
NM_080826.1 XM_017027680.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4096697 c.896G>A p.R299H Chr.20: 13298960-13298960 on Build GRCh38
1410416 c.906C>T p.S302S Chr.20: 13298970-13298970 on Build GRCh38
2703214 c.899G>A p.W300* Chr.20: 13298963-13298963 on Build GRCh38
4693181 c.878-1G>A p.? Chr.20: 13298941-13298941 on Build GRCh38
1285587 c.881C>A p.T294K Chr.20: 13298945-13298945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148388492 Chr.20: 13298866-13298866 on Build GRCh38
rs547722323 Chr.20: 13298773-13298773 on Build GRCh38
rs1027169564 Chr.20: 13298871-13298871 on Build GRCh38
rs753397350 Chr.20: 13298744-13298744 on Build GRCh38
rs778504431 Chr.20: 13298926-13298926 on Build GRCh38
rs538827724 Chr.20: 13298849-13298849 on Build GRCh38
rs561234856 Chr.20: 13298792-13298792 on Build GRCh38
rs959218819 Chr.20: 13298836-13298836 on Build GRCh38
rs989433597 Chr.20: 13298960-13298960 on Build GRCh38
rs6078980 Chr.20: 13298764-13298764 on Build GRCh38
rs918605437 Chr.20: 13298862-13298862 on Build GRCh38
rs770049999 Chr.20: 13298956-13298956 on Build GRCh38
rs530173673 Chr.20: 13298809-13298809 on Build GRCh38
rs368477696 Chr.20: 13298779-13298779 on Build GRCh38
rs76135955 Chr.20: 13298759-13298759 on Build GRCh38
rs371226530 Chr.20: 13298932-13298932 on Build GRCh38
rs779369275 Chr.20: 13298816-13298816 on Build GRCh38
rs921218476 Chr.20: 13298761-13298761 on Build GRCh38
rs927901631 Chr.20: 13298821-13298821 on Build GRCh38
rs574471897 Chr.20: 13298919-13298919 on Build GRCh38
rs1036954434 Chr.20: 13298852-13298852 on Build GRCh38
rs938685553 Chr.20: 13298844-13298844 on Build GRCh38
rs1034940385 Chr.20: 13298799-13298815 on Build GRCh38
rs763009893 Chr.20: 13298892-13298892 on Build GRCh38
rs554517627 Chr.20: 13298910-13298910 on Build GRCh38
rs965095330 Chr.20: 13298905-13298905 on Build GRCh38
rs141402001 Chr.20: 13298899-13298899 on Build GRCh38
rs1041838438 Chr.20: 13298840-13298840 on Build GRCh38
rs975065353 Chr.20: 13298743-13298743 on Build GRCh38
rs764630816 Chr.20: 13298895-13298895 on Build GRCh38
rs982066969 Chr.20: 13298808-13298808 on Build GRCh38
rs777651612 Chr.20: 13298936-13298936 on Build GRCh38
rs890056411 Chr.20: 13298894-13298894 on Build GRCh38
rs750802692 Chr.20: 13298909-13298909 on Build GRCh38
rs957810728 Chr.20: 13298755-13298755 on Build GRCh38
rs556763641 Chr.20: 13298946-13298946 on Build GRCh38
rs754491572 Chr.20: 13298925-13298925 on Build GRCh38
rs894753550 Chr.20: 13298851-13298851 on Build GRCh38
rs189481232 Chr.20: 13298908-13298908 on Build GRCh38
rs766016612 Chr.20: 13298911-13298911 on Build GRCh38
rs752638397 Chr.20: 13298931-13298931 on Build GRCh38
rs1019324333 Chr.20: 13298900-13298900 on Build GRCh38
rs116494408 Chr.20: 13298843-13298843 on Build GRCh38
rs991101102 Chr.20: 13298839-13298839 on Build GRCh38
rs549729878 Chr.20: 13298814-13298814 on Build GRCh38

View Full Product Details