Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 20749563-20749836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNIP4
Entrez Gene ID
80333
Gene Name
potassium voltage-gated channel interacting protein 4
Gene Aliases
CALP, KCHIP4
UniGene
Hs.655705
Gene Chromosome Location
Chr.4: 20728520-21948801 on Build GRCh38
Transcripts
NM_001035003.1 NM_001035004.1 NM_025221.5 NM_147181.3 NM_147182.3 NM_147183.3 XM_005248190.4 XM_011513882.2 XM_011513883.2 XM_011513884.2 XM_011513885.2 XM_011513886.2 XM_011513887.2 XM_011513889.2 XM_017008653.1 NR_133933.1

Gene Symbol
PACRGL
Entrez Gene ID
133015
Gene Name
PARK2 coregulated like
Gene Aliases
C4orf28
UniGene
Hs.479298
Gene Chromosome Location
Chr.4: 20696282-20757766 on Build GRCh38
Transcripts
NM_001035003.1 NM_001035004.1 NM_025221.5 NM_147181.3 NM_147182.3 NM_147183.3 XM_005248190.4 XM_011513882.2 XM_011513883.2 XM_011513884.2 XM_011513885.2 XM_011513886.2 XM_011513887.2 XM_011513889.2 XM_017008653.1 NR_133933.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3132626 c.337A>G p.T113A Chr.4: 20749691-20749691 on Build GRCh38
3373467 c.310T>G p.F104V Chr.4: 20749706-20749706 on Build GRCh38
7401404 c.397G>A p.D133N Chr.4: 20749694-20749694 on Build GRCh38
5060305 c.354T>C p.A118A Chr.4: 20749674-20749674 on Build GRCh38
1753682 c.420G>A p.V140V Chr.4: 20749671-20749671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780303241 Chr.4: 20749613-20749613 on Build GRCh38
rs2322688 Chr.4: 20749655-20749655 on Build GRCh38
rs777091293 Chr.4: 20749653-20749653 on Build GRCh38
rs756779365 Chr.4: 20749680-20749680 on Build GRCh38
rs374097995 Chr.4: 20749741-20749741 on Build GRCh38
rs772509553 Chr.4: 20749627-20749627 on Build GRCh38
rs750337453 Chr.4: 20749691-20749691 on Build GRCh38
rs112653243 Chr.4: 20749783-20749783 on Build GRCh38
rs932421345 Chr.4: 20749768-20749768 on Build GRCh38
rs749012604 Chr.4: 20749722-20749722 on Build GRCh38
rs73802309 Chr.4: 20749597-20749597 on Build GRCh38
rs768879714 Chr.4: 20749615-20749615 on Build GRCh38
rs767393802 Chr.4: 20749681-20749681 on Build GRCh38
rs748240446 Chr.4: 20749764-20749764 on Build GRCh38
rs763735612 Chr.4: 20749656-20749656 on Build GRCh38
rs779855024 Chr.4: 20749719-20749719 on Build GRCh38
rs370308743 Chr.4: 20749636-20749636 on Build GRCh38
rs1049471516 Chr.4: 20749776-20749776 on Build GRCh38
rs757747371 Chr.4: 20749798-20749798 on Build GRCh38
rs761164144 Chr.4: 20749641-20749641 on Build GRCh38
rs779180133 Chr.4: 20749757-20749757 on Build GRCh38
rs755996521 Chr.4: 20749706-20749706 on Build GRCh38
rs772163502 Chr.4: 20749781-20749781 on Build GRCh38
rs774295683 Chr.4: 20749616-20749616 on Build GRCh38
rs773853961 Chr.4: 20749635-20749635 on Build GRCh38
rs922388861 Chr.4: 20749743-20749743 on Build GRCh38
rs369759324 Chr.4: 20749608-20749608 on Build GRCh38
rs751047200 Chr.4: 20749665-20749665 on Build GRCh38
rs949696688 Chr.4: 20749807-20749807 on Build GRCh38
rs749534148 Chr.4: 20749614-20749614 on Build GRCh38
rs748198574 Chr.4: 20749623-20749623 on Build GRCh38
rs953784613 Chr.4: 20749740-20749740 on Build GRCh38
rs766832653 Chr.4: 20749644-20749644 on Build GRCh38
rs764506120 Chr.4: 20749772-20749772 on Build GRCh38

View Full Product Details