Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23254096-23254355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPNMB
Entrez Gene ID
10457
Gene Name
glycoprotein nmb
Gene Aliases
HGFIN, NMB
UniGene
Hs.190495
Gene Chromosome Location
Chr.7: 23246206-23275110 on Build GRCh38
Transcripts
NM_001005340.1 NM_002510.2 XM_005249578.2 XM_017011676.1 XM_017011677.1 XM_017011678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3026140 c.306C>T p.F102F Chr.7: 23254251-23254251 on Build GRCh38
3637513 c.252T>C p.S84S Chr.7: 23254197-23254197 on Build GRCh38
3026141 c.344T>A p.V115D Chr.7: 23254289-23254289 on Build GRCh38
3663112 c.323A>G p.E108G Chr.7: 23254268-23254268 on Build GRCh38
7265039 c.367+3A>C p.? Chr.7: 23254315-23254315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749959364 Chr.7: 23254256-23254256 on Build GRCh38
rs879123746 Chr.7: 23254291-23254291 on Build GRCh38
rs533327850 Chr.7: 23254208-23254208 on Build GRCh38
rs772625202 Chr.7: 23254142-23254142 on Build GRCh38
rs370724348 Chr.7: 23254278-23254278 on Build GRCh38
rs183275111 Chr.7: 23254165-23254165 on Build GRCh38
rs979606166 Chr.7: 23254251-23254251 on Build GRCh38
rs748648952 Chr.7: 23254132-23254132 on Build GRCh38
rs369065423 Chr.7: 23254164-23254164 on Build GRCh38
rs745549323 Chr.7: 23254175-23254175 on Build GRCh38
rs113932410 Chr.7: 23254212-23254212 on Build GRCh38
rs772283661 Chr.7: 23254154-23254154 on Build GRCh38
rs199347 Chr.7: 23254127-23254127 on Build GRCh38
rs773200861 Chr.7: 23254145-23254145 on Build GRCh38
rs755230681 Chr.7: 23254176-23254176 on Build GRCh38
rs373987305 Chr.7: 23254314-23254314 on Build GRCh38
rs571001843 Chr.7: 23254202-23254202 on Build GRCh38
rs551095019 Chr.7: 23254194-23254194 on Build GRCh38
rs145407985 Chr.7: 23254184-23254184 on Build GRCh38
rs758729806 Chr.7: 23254255-23254268 on Build GRCh38
rs1043296124 Chr.7: 23254160-23254160 on Build GRCh38
rs200247677 Chr.7: 23254171-23254171 on Build GRCh38
rs150027317 Chr.7: 23254169-23254169 on Build GRCh38
rs142450741 Chr.7: 23254213-23254213 on Build GRCh38
rs751571747 Chr.7: 23254236-23254236 on Build GRCh38
rs776647076 Chr.7: 23254207-23254207 on Build GRCh38
rs375459434 Chr.7: 23254320-23254320 on Build GRCh38
rs973959143 Chr.7: 23254205-23254205 on Build GRCh38
rs776930697 Chr.7: 23254161-23254161 on Build GRCh38
rs755767733 Chr.7: 23254274-23254274 on Build GRCh38
rs765081388 Chr.7: 23254163-23254163 on Build GRCh38
rs377653957 Chr.7: 23254316-23254316 on Build GRCh38
rs756824456 Chr.7: 23254173-23254173 on Build GRCh38
rs186933275 Chr.7: 23254279-23254279 on Build GRCh38
rs775407617 Chr.7: 23254215-23254215 on Build GRCh38
rs149206943 Chr.7: 23254185-23254185 on Build GRCh38
rs199346 Chr.7: 23254153-23254153 on Build GRCh38
rs767058218 Chr.7: 23254243-23254243 on Build GRCh38
rs202078752 Chr.7: 23254235-23254235 on Build GRCh38
rs766470213 Chr.7: 23254150-23254150 on Build GRCh38
rs199988275 Chr.7: 23254288-23254288 on Build GRCh38
rs867127640 Chr.7: 23254229-23254229 on Build GRCh38
rs747009853 Chr.7: 23254196-23254196 on Build GRCh38
rs756663981 Chr.7: 23254237-23254237 on Build GRCh38
rs745988149 Chr.7: 23254327-23254327 on Build GRCh38
rs747552463 Chr.7: 23254317-23254317 on Build GRCh38
rs200768193 Chr.7: 23254286-23254286 on Build GRCh38
rs759191333 Chr.7: 23254162-23254162 on Build GRCh38
rs370920664 Chr.7: 23254319-23254319 on Build GRCh38
rs773737053 Chr.7: 23254188-23254188 on Build GRCh38
rs761061706 Chr.7: 23254288-23254293 on Build GRCh38
rs200133864 Chr.7: 23254144-23254144 on Build GRCh38
rs964181513 Chr.7: 23254249-23254249 on Build GRCh38
rs779235718 Chr.7: 23254178-23254178 on Build GRCh38
rs372684901 Chr.7: 23254174-23254174 on Build GRCh38

View Full Product Details