Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26032981-26033254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H2AB
Entrez Gene ID
8335
Gene Name
histone cluster 1, H2ab
Gene Aliases
H2A/m, H2AFM
UniGene
Hs.248174
Gene Chromosome Location
Chr.6: 26033092-26033568 on Build GRCh38
Transcripts
NM_003513.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6680219 c.331A>G p.N111D Chr.6: 26033238-26033238 on Build GRCh38
4852428 c.392G>A p.*131* Chr.6: 26033177-26033177 on Build GRCh38
5421822 c.378G>C p.K126N Chr.6: 26033191-26033191 on Build GRCh38
450998 c.364G>C p.E122Q Chr.6: 26033205-26033205 on Build GRCh38
6680220 c.369C>T p.S123S Chr.6: 26033200-26033200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112700668 Chr.6: 26033018-26033018 on Build GRCh38
rs749260550 Chr.6: 26033224-26033224 on Build GRCh38
rs757022005 Chr.6: 26033199-26033199 on Build GRCh38
rs199586241 Chr.6: 26033189-26033189 on Build GRCh38
rs1006915413 Chr.6: 26033207-26033207 on Build GRCh38
rs974745516 Chr.6: 26033012-26033012 on Build GRCh38
rs769832299 Chr.6: 26033162-26033162 on Build GRCh38
rs985288793 Chr.6: 26033135-26033135 on Build GRCh38
rs746136514 Chr.6: 26033158-26033158 on Build GRCh38
rs760712565 Chr.6: 26033201-26033202 on Build GRCh38
rs752086646 Chr.6: 26033191-26033191 on Build GRCh38
rs752653377 Chr.6: 26033147-26033147 on Build GRCh38
rs991804863 Chr.6: 26033050-26033050 on Build GRCh38
rs917707681 Chr.6: 26033051-26033051 on Build GRCh38
rs374459306 Chr.6: 26033031-26033031 on Build GRCh38
rs777775837 Chr.6: 26033153-26033153 on Build GRCh38
rs955876241 Chr.6: 26033095-26033095 on Build GRCh38
rs533561365 Chr.6: 26033015-26033015 on Build GRCh38
rs997865488 Chr.6: 26033111-26033111 on Build GRCh38
rs867445538 Chr.6: 26033053-26033053 on Build GRCh38
rs745693623 Chr.6: 26033205-26033205 on Build GRCh38
rs777564596 Chr.6: 26033195-26033195 on Build GRCh38
rs150171580 Chr.6: 26033157-26033157 on Build GRCh38
rs980625696 Chr.6: 26033063-26033063 on Build GRCh38
rs895400176 Chr.6: 26033028-26033028 on Build GRCh38
rs1043904455 Chr.6: 26033082-26033082 on Build GRCh38
rs771905911 Chr.6: 26033187-26033191 on Build GRCh38
rs904264757 Chr.6: 26033086-26033086 on Build GRCh38
rs918696261 Chr.6: 26033217-26033217 on Build GRCh38
rs148098876 Chr.6: 26033008-26033008 on Build GRCh38
rs751553290 Chr.6: 26033196-26033196 on Build GRCh38
rs377552879 Chr.6: 26033133-26033133 on Build GRCh38
rs550846028 Chr.6: 26033227-26033227 on Build GRCh38
rs56321056 Chr.6: 26033133-26033134 on Build GRCh38
rs556828456 Chr.6: 26033119-26033119 on Build GRCh38
rs368605549 Chr.6: 26033060-26033060 on Build GRCh38
rs768055353 Chr.6: 26033185-26033185 on Build GRCh38
rs775391103 Chr.6: 26033164-26033164 on Build GRCh38
rs757926888 Chr.6: 26033194-26033194 on Build GRCh38
rs774432400 Chr.6: 26033067-26033067 on Build GRCh38
rs778619218 Chr.6: 26033146-26033146 on Build GRCh38
rs773101337 Chr.6: 26033191-26033192 on Build GRCh38
rs766063322 Chr.6: 26033136-26033136 on Build GRCh38
rs1031434353 Chr.6: 26033092-26033092 on Build GRCh38
rs762265818 Chr.6: 26033184-26033184 on Build GRCh38
rs774927927 Chr.6: 26033183-26033183 on Build GRCh38
rs576786401 Chr.6: 26033120-26033120 on Build GRCh38
rs763116503 Chr.6: 26033172-26033172 on Build GRCh38
rs768986746 Chr.6: 26033174-26033174 on Build GRCh38
rs753614523 Chr.6: 26033145-26033145 on Build GRCh38
rs753522467 Chr.6: 26033188-26033188 on Build GRCh38
rs758233079 Chr.6: 26033152-26033152 on Build GRCh38
rs751822392 Chr.6: 26033134-26033134 on Build GRCh38
rs111714517 Chr.6: 26033126-26033126 on Build GRCh38
rs766399143 Chr.6: 26033205-26033206 on Build GRCh38
rs769954431 Chr.6: 26033212-26033212 on Build GRCh38
rs780337319 Chr.6: 26033220-26033220 on Build GRCh38
rs746711691 Chr.6: 26033156-26033156 on Build GRCh38
rs767341049 Chr.6: 26033214-26033215 on Build GRCh38
rs999933281 Chr.6: 26033088-26033088 on Build GRCh38
rs761312976 Chr.6: 26033151-26033153 on Build GRCh38
rs553782455 Chr.6: 26033017-26033017 on Build GRCh38
rs137942115 Chr.6: 26033190-26033190 on Build GRCh38
rs781004178 Chr.6: 26033202-26033202 on Build GRCh38

View Full Product Details