Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15115411-15115684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILVBL
Entrez Gene ID
10994
Gene Name
ilvB acetolactate synthase like
Gene Aliases
209L8, AHAS, HACL1L, ILV2H
UniGene
Hs.78880
Gene Chromosome Location
Chr.19: 15114974-15125799 on Build GRCh38
Transcripts
NM_006844.4 XM_005259717.4 XM_011527651.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146193 c.1615-2A>G p.? Chr.19: 15115673-15115673 on Build GRCh38
5497297 c.1721-1G>A p.? Chr.19: 15115431-15115431 on Build GRCh38
7205516 c.1615-2A>C p.? Chr.19: 15115673-15115673 on Build GRCh38
1391032 c.1696G>A p.V566M Chr.19: 15115590-15115590 on Build GRCh38
5365322 c.1707C>T p.G569G Chr.19: 15115579-15115579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750694560 Chr.19: 15115476-15115476 on Build GRCh38
rs773383279 Chr.19: 15115591-15115591 on Build GRCh38
rs530673298 Chr.19: 15115617-15115617 on Build GRCh38
rs759101724 Chr.19: 15115448-15115448 on Build GRCh38
rs774333276 Chr.19: 15115569-15115569 on Build GRCh38
rs562012937 Chr.19: 15115520-15115525 on Build GRCh38
rs745458168 Chr.19: 15115644-15115644 on Build GRCh38
rs935576273 Chr.19: 15115502-15115502 on Build GRCh38
rs560867058 Chr.19: 15115530-15115530 on Build GRCh38
rs1020393869 Chr.19: 15115640-15115640 on Build GRCh38
rs779643093 Chr.19: 15115657-15115657 on Build GRCh38
rs1007108019 Chr.19: 15115549-15115549 on Build GRCh38
rs374158124 Chr.19: 15115620-15115620 on Build GRCh38
rs765825330 Chr.19: 15115619-15115619 on Build GRCh38
rs373857711 Chr.19: 15115510-15115510 on Build GRCh38
rs776491990 Chr.19: 15115444-15115444 on Build GRCh38
rs760981000 Chr.19: 15115592-15115592 on Build GRCh38
rs755522690 Chr.19: 15115519-15115519 on Build GRCh38
rs112758281 Chr.19: 15115450-15115450 on Build GRCh38
rs780214868 Chr.19: 15115478-15115478 on Build GRCh38
rs748820110 Chr.19: 15115658-15115658 on Build GRCh38
rs542320993 Chr.19: 15115515-15115515 on Build GRCh38
rs377592260 Chr.19: 15115477-15115477 on Build GRCh38
rs754250466 Chr.19: 15115517-15115517 on Build GRCh38
rs763743114 Chr.19: 15115453-15115453 on Build GRCh38
rs767715397 Chr.19: 15115474-15115474 on Build GRCh38
rs755077380 Chr.19: 15115613-15115613 on Build GRCh38
rs753956935 Chr.19: 15115602-15115602 on Build GRCh38
rs749118182 Chr.19: 15115558-15115558 on Build GRCh38
rs531809801 Chr.19: 15115470-15115482 on Build GRCh38
rs941375041 Chr.19: 15115458-15115458 on Build GRCh38
rs373878440 Chr.19: 15115451-15115451 on Build GRCh38
rs755799977 Chr.19: 15115656-15115656 on Build GRCh38
rs780966130 Chr.19: 15115545-15115545 on Build GRCh38
rs770710688 Chr.19: 15115442-15115442 on Build GRCh38
rs370255199 Chr.19: 15115523-15115523 on Build GRCh38
rs767342989 Chr.19: 15115590-15115590 on Build GRCh38
rs761575531 Chr.19: 15115579-15115579 on Build GRCh38
rs368645720 Chr.19: 15115551-15115551 on Build GRCh38
rs143219140 Chr.19: 15115630-15115630 on Build GRCh38
rs200352922 Chr.19: 15115536-15115536 on Build GRCh38
rs745552086 Chr.19: 15115550-15115550 on Build GRCh38
rs778308860 Chr.19: 15115641-15115641 on Build GRCh38
rs1014822219 Chr.19: 15115632-15115632 on Build GRCh38
rs763247904 Chr.19: 15115627-15115628 on Build GRCh38
rs1029661687 Chr.19: 15115595-15115595 on Build GRCh38
rs371516271 Chr.19: 15115563-15115563 on Build GRCh38
rs761667265 Chr.19: 15115466-15115466 on Build GRCh38

View Full Product Details