Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18434824-18435085 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISYNA1
Entrez Gene ID
51477
Gene Name
inositol-3-phosphate synthase 1
Gene Aliases
INO1, INOS, IPS, IPS 1, IPS-1
UniGene
Hs.405873
Gene Chromosome Location
Chr.19: 18434388-18438301 on Build GRCh38
Transcripts
NM_001170938.1 NM_001253389.1 NM_016368.4 NR_045573.1 NR_045574.1 XM_011528059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4693194 c.1598C>T p.P533L Chr.19: 18434992-18434992 on Build GRCh38
993093 c.1599G>A p.P533P Chr.19: 18434991-18434991 on Build GRCh38
3937945 c.1661C>G p.P554R Chr.19: 18434929-18434929 on Build GRCh38
4922080 c.1626C>A p.T542T Chr.19: 18434964-18434964 on Build GRCh38
3100032 c.1602A>G p.V534V Chr.19: 18434988-18434988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774163346 Chr.19: 18435063-18435063 on Build GRCh38
rs746430923 Chr.19: 18434876-18434876 on Build GRCh38
rs749954642 Chr.19: 18435010-18435010 on Build GRCh38
rs757930650 Chr.19: 18434936-18434936 on Build GRCh38
rs539012860 Chr.19: 18435057-18435057 on Build GRCh38
rs987418775 Chr.19: 18434962-18434962 on Build GRCh38
rs775082005 Chr.19: 18434985-18434985 on Build GRCh38
rs372353337 Chr.19: 18435062-18435062 on Build GRCh38
rs756745983 Chr.19: 18434957-18434957 on Build GRCh38
rs570793925 Chr.19: 18435032-18435032 on Build GRCh38
rs536858921 Chr.19: 18434991-18434991 on Build GRCh38
rs753774437 Chr.19: 18434898-18434899 on Build GRCh38
rs954500894 Chr.19: 18434949-18434949 on Build GRCh38
rs768941131 Chr.19: 18434894-18434894 on Build GRCh38
rs746859151 Chr.19: 18434975-18434977 on Build GRCh38
rs762837471 Chr.19: 18434873-18434873 on Build GRCh38
rs201744334 Chr.19: 18434907-18434907 on Build GRCh38
rs149546342 Chr.19: 18434937-18434937 on Build GRCh38
rs748464658 Chr.19: 18434950-18434950 on Build GRCh38
rs200221382 Chr.19: 18435009-18435009 on Build GRCh38
rs752559696 Chr.19: 18434933-18434933 on Build GRCh38
rs35132827 Chr.19: 18435047-18435047 on Build GRCh38
rs763042538 Chr.19: 18434891-18434891 on Build GRCh38
rs780728546 Chr.19: 18434958-18434958 on Build GRCh38
rs766973409 Chr.19: 18435000-18435000 on Build GRCh38
rs1045631 Chr.19: 18434976-18434976 on Build GRCh38
rs3185558 Chr.19: 18434893-18434893 on Build GRCh38
rs1003258871 Chr.19: 18435019-18435019 on Build GRCh38
rs781603897 Chr.19: 18434997-18434997 on Build GRCh38
rs774704289 Chr.19: 18434967-18434967 on Build GRCh38
rs774234744 Chr.19: 18434897-18434897 on Build GRCh38
rs1045641 Chr.19: 18434945-18434945 on Build GRCh38
rs773367547 Chr.19: 18435049-18435049 on Build GRCh38
rs772449312 Chr.19: 18435037-18435037 on Build GRCh38
rs147275395 Chr.19: 18435029-18435029 on Build GRCh38
rs758069540 Chr.19: 18434929-18434930 on Build GRCh38
rs141254112 Chr.19: 18435021-18435021 on Build GRCh38
rs758811983 Chr.19: 18434963-18434963 on Build GRCh38
rs778673490 Chr.19: 18434898-18434901 on Build GRCh38
rs930950192 Chr.19: 18434995-18434995 on Build GRCh38
rs771274606 Chr.19: 18434996-18434999 on Build GRCh38
rs759499204 Chr.19: 18434982-18434982 on Build GRCh38
rs148355784 Chr.19: 18434880-18434880 on Build GRCh38
rs768690400 Chr.19: 18434965-18434965 on Build GRCh38
rs761985931 Chr.19: 18434906-18434906 on Build GRCh38
rs775486515 Chr.19: 18435061-18435061 on Build GRCh38
rs903817483 Chr.19: 18434899-18434899 on Build GRCh38
rs751189492 Chr.19: 18434992-18434992 on Build GRCh38
rs748432974 Chr.19: 18434968-18434968 on Build GRCh38
rs201345070 Chr.19: 18434890-18434890 on Build GRCh38
rs980191950 Chr.19: 18434996-18434996 on Build GRCh38
rs765234659 Chr.19: 18434984-18434984 on Build GRCh38
rs746537378 Chr.19: 18434867-18434867 on Build GRCh38
rs751281429 Chr.19: 18434951-18434951 on Build GRCh38
rs764904770 Chr.19: 18434923-18434923 on Build GRCh38
rs544498087 Chr.19: 18434865-18434865 on Build GRCh38
rs777714813 Chr.19: 18434960-18434967 on Build GRCh38
rs755350113 Chr.19: 18435023-18435023 on Build GRCh38
rs143175757 Chr.19: 18434974-18434974 on Build GRCh38
rs564672040 Chr.19: 18434964-18434964 on Build GRCh38
rs747117719 Chr.19: 18435056-18435056 on Build GRCh38
rs563687209 Chr.19: 18434856-18434856 on Build GRCh38
rs1063525 Chr.19: 18434921-18434921 on Build GRCh38
rs776690343 Chr.19: 18434920-18434920 on Build GRCh38
rs972218642 Chr.19: 18434917-18434917 on Build GRCh38
rs367687629 Chr.19: 18434868-18434868 on Build GRCh38
rs143059785 Chr.19: 18434846-18434846 on Build GRCh38
rs972918430 Chr.19: 18434981-18434981 on Build GRCh38
rs772251805 Chr.19: 18434971-18434971 on Build GRCh38
rs762804918 Chr.19: 18434990-18434990 on Build GRCh38
rs747312137 Chr.19: 18435016-18435016 on Build GRCh38
rs1036329677 Chr.19: 18434870-18434870 on Build GRCh38
rs986928969 Chr.19: 18434966-18434966 on Build GRCh38

View Full Product Details