Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18059753-18060024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL12RB1
Entrez Gene ID
3594
Gene Name
interleukin 12 receptor subunit beta 1
Gene Aliases
CD212, IL-12R-BETA1, IL12RB, IMD30
UniGene
Hs.567294
Gene Chromosome Location
Chr.19: 18058994-18099027 on Build GRCh38
Transcripts
NM_001290023.1 NM_001290024.1 NM_005535.2 XM_006722741.3 XM_011527966.2 XM_011527967.2 XM_011527968.2 XM_011527969.2 XM_011527970.2 XM_011527971.2 XM_011527972.2 XM_011527973.2 XM_011527974.2 XM_011527975.2 XM_017026762.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7220394 c.1878C>T p.G626G Chr.19: 18059999-18059999 on Build GRCh38
6557646 c.1923C>A p.A641A Chr.19: 18059954-18059954 on Build GRCh38
3530662 c.1876G>A p.G626S Chr.19: 18060001-18060001 on Build GRCh38
4474459 c.1922C>T p.A641V Chr.19: 18059955-18059955 on Build GRCh38
6084203 c.1923C>T p.A641A Chr.19: 18059954-18059954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003563647 Chr.19: 18059937-18059937 on Build GRCh38
rs776875362 Chr.19: 18059933-18059934 on Build GRCh38
rs748085048 Chr.19: 18059821-18059821 on Build GRCh38
rs566576601 Chr.19: 18059905-18059905 on Build GRCh38
rs761603497 Chr.19: 18059884-18059884 on Build GRCh38
rs1017464476 Chr.19: 18059795-18059795 on Build GRCh38
rs955091939 Chr.19: 18059833-18059833 on Build GRCh38
rs748543103 Chr.19: 18059834-18059834 on Build GRCh38
rs12461312 Chr.19: 18059847-18059847 on Build GRCh38
rs970598928 Chr.19: 18059842-18059842 on Build GRCh38
rs17882555 Chr.19: 18059870-18059870 on Build GRCh38
rs143367415 Chr.19: 18059998-18059998 on Build GRCh38
rs772340282 Chr.19: 18059980-18059980 on Build GRCh38
rs775529244 Chr.19: 18059985-18059985 on Build GRCh38
rs1005627741 Chr.19: 18059781-18059781 on Build GRCh38
rs868145882 Chr.19: 18059914-18059914 on Build GRCh38
rs376525105 Chr.19: 18059869-18059869 on Build GRCh38
rs775231157 Chr.19: 18059866-18059866 on Build GRCh38
rs769424178 Chr.19: 18059981-18059981 on Build GRCh38
rs894458142 Chr.19: 18059799-18059799 on Build GRCh38
rs750881845 Chr.19: 18059913-18059913 on Build GRCh38
rs865848042 Chr.19: 18059921-18059921 on Build GRCh38
rs761145805 Chr.19: 18060000-18060000 on Build GRCh38
rs754254406 Chr.19: 18059939-18059939 on Build GRCh38
rs866267146 Chr.19: 18059907-18059907 on Build GRCh38
rs755120896 Chr.19: 18059953-18059953 on Build GRCh38
rs751046730 Chr.19: 18059879-18059879 on Build GRCh38
rs759314886 Chr.19: 18059864-18059864 on Build GRCh38
rs772945592 Chr.19: 18059999-18059999 on Build GRCh38
rs750790051 Chr.19: 18059844-18059844 on Build GRCh38
rs370240272 Chr.19: 18059848-18059848 on Build GRCh38
rs1024323957 Chr.19: 18059840-18059840 on Build GRCh38
rs553163198 Chr.19: 18059875-18059875 on Build GRCh38
rs768612090 Chr.19: 18059990-18059990 on Build GRCh38
rs773786998 Chr.19: 18059994-18059994 on Build GRCh38
rs745932745 Chr.19: 18059882-18059882 on Build GRCh38
rs777955828 Chr.19: 18059843-18059843 on Build GRCh38
rs202106699 Chr.19: 18059917-18059917 on Build GRCh38
rs146334638 Chr.19: 18059864-18059864 on Build GRCh38
rs769726124 Chr.19: 18059865-18059865 on Build GRCh38
rs972995442 Chr.19: 18059785-18059785 on Build GRCh38
rs776991418 Chr.19: 18059854-18059854 on Build GRCh38
rs748205033 Chr.19: 18059969-18059969 on Build GRCh38
rs779222722 Chr.19: 18059858-18059858 on Build GRCh38
rs748330114 Chr.19: 18059862-18059862 on Build GRCh38
rs1055504589 Chr.19: 18059955-18059957 on Build GRCh38
rs780627570 Chr.19: 18059933-18059933 on Build GRCh38
rs199686420 Chr.19: 18059963-18059963 on Build GRCh38
rs776461067 Chr.19: 18059863-18059863 on Build GRCh38
rs761401659 Chr.19: 18059996-18059996 on Build GRCh38
rs756740790 Chr.19: 18059845-18059845 on Build GRCh38
rs758963566 Chr.19: 18059838-18059838 on Build GRCh38
rs1054291798 Chr.19: 18059859-18059860 on Build GRCh38

View Full Product Details