Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 220102390-220102635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS2
Entrez Gene ID
55699
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
Gene Aliases
CAGSSS, ILERS
UniGene
Hs.262823
Gene Chromosome Location
Chr.1: 220094113-220148041 on Build GRCh38
Transcripts
NM_018060.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6865204 c.730T>G p.F244V Chr.1: 220102393-220102393 on Build GRCh38
1959100 c.749+2T>C p.? Chr.1: 220102414-220102414 on Build GRCh38
1338968 c.808T>C p.Y270H Chr.1: 220102553-220102553 on Build GRCh38
6865209 c.729G>T p.V243V Chr.1: 220102392-220102392 on Build GRCh38
1210208 c.785C>A p.P262H Chr.1: 220102530-220102530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201916678 Chr.1: 220102540-220102540 on Build GRCh38
rs746455426 Chr.1: 220102533-220102533 on Build GRCh38
rs377432411 Chr.1: 220102545-220102545 on Build GRCh38
rs958850547 Chr.1: 220102443-220102443 on Build GRCh38
rs774960759 Chr.1: 220102463-220102463 on Build GRCh38
rs766651661 Chr.1: 220102493-220102493 on Build GRCh38
rs762202072 Chr.1: 220102465-220102465 on Build GRCh38
rs769112693 Chr.1: 220102553-220102553 on Build GRCh38
rs762450476 Chr.1: 220102592-220102592 on Build GRCh38
rs756410015 Chr.1: 220102490-220102490 on Build GRCh38
rs774564592 Chr.1: 220102566-220102566 on Build GRCh38
rs917298497 Chr.1: 220102544-220102544 on Build GRCh38
rs751095095 Chr.1: 220102512-220102512 on Build GRCh38
rs372618006 Chr.1: 220102429-220102429 on Build GRCh38
rs370306595 Chr.1: 220102452-220102452 on Build GRCh38
rs904984929 Chr.1: 220102455-220102455 on Build GRCh38
rs760791870 Chr.1: 220102602-220102602 on Build GRCh38
rs776306373 Chr.1: 220102537-220102537 on Build GRCh38
rs1032292776 Chr.1: 220102518-220102518 on Build GRCh38
rs778544749 Chr.1: 220102507-220102507 on Build GRCh38
rs201539020 Chr.1: 220102436-220102436 on Build GRCh38
rs777266427 Chr.1: 220102531-220102531 on Build GRCh38
rs772640835 Chr.1: 220102593-220102593 on Build GRCh38
rs754766431 Chr.1: 220102498-220102498 on Build GRCh38
rs764539319 Chr.1: 220102449-220102449 on Build GRCh38
rs750528193 Chr.1: 220102478-220102478 on Build GRCh38
rs767766570 Chr.1: 220102467-220102467 on Build GRCh38
rs1009545961 Chr.1: 220102536-220102536 on Build GRCh38
rs772692964 Chr.1: 220102423-220102423 on Build GRCh38
rs747645857 Chr.1: 220102420-220102420 on Build GRCh38
rs374904870 Chr.1: 220102430-220102430 on Build GRCh38
rs758325882 Chr.1: 220102529-220102529 on Build GRCh38
rs370222815 Chr.1: 220102438-220102438 on Build GRCh38
rs373433981 Chr.1: 220102520-220102520 on Build GRCh38
rs753601921 Chr.1: 220102497-220102497 on Build GRCh38
rs770588599 Chr.1: 220102534-220102534 on Build GRCh38
rs976770107 Chr.1: 220102591-220102591 on Build GRCh38
rs773439017 Chr.1: 220102594-220102594 on Build GRCh38
rs775841640 Chr.1: 220102431-220102431 on Build GRCh38

View Full Product Details