Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138181274-138181545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF20A
Entrez Gene ID
10112
Gene Name
kinesin family member 20A
Gene Aliases
MKLP2, RAB6KIFL
UniGene
Hs.718626
Gene Chromosome Location
Chr.5: 138178728-138187715 on Build GRCh38
Transcripts
NM_005733.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1061249 c.204C>A p.Y68* Chr.5: 138181460-138181460 on Build GRCh38
4971066 c.170C>T p.P57L Chr.5: 138181426-138181426 on Build GRCh38
5727688 c.242G>A p.R81Q Chr.5: 138181498-138181498 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs943318032 Chr.5: 138181397-138181397 on Build GRCh38
rs998821911 Chr.5: 138181355-138181355 on Build GRCh38
rs776713008 Chr.5: 138181380-138181380 on Build GRCh38
rs115502334 Chr.5: 138181316-138181316 on Build GRCh38
rs766950655 Chr.5: 138181478-138181478 on Build GRCh38
rs527740679 Chr.5: 138181498-138181498 on Build GRCh38
rs751203875 Chr.5: 138181431-138181431 on Build GRCh38
rs763706536 Chr.5: 138181422-138181422 on Build GRCh38
rs902077802 Chr.5: 138181416-138181416 on Build GRCh38
rs755397908 Chr.5: 138181493-138181493 on Build GRCh38
rs146894462 Chr.5: 138181448-138181448 on Build GRCh38
rs762779391 Chr.5: 138181417-138181417 on Build GRCh38
rs771127886 Chr.5: 138181379-138181379 on Build GRCh38
rs1018345097 Chr.5: 138181335-138181335 on Build GRCh38
rs758776117 Chr.5: 138181503-138181503 on Build GRCh38
rs771147897 Chr.5: 138181515-138181515 on Build GRCh38
rs1006665435 Chr.5: 138181327-138181328 on Build GRCh38
rs998204951 Chr.5: 138181429-138181429 on Build GRCh38
rs3734116 Chr.5: 138181443-138181443 on Build GRCh38
rs893228942 Chr.5: 138181376-138181376 on Build GRCh38
rs865809971 Chr.5: 138181326-138181326 on Build GRCh38
rs779440603 Chr.5: 138181497-138181497 on Build GRCh38
rs372749091 Chr.5: 138181473-138181473 on Build GRCh38
rs778206749 Chr.5: 138181505-138181505 on Build GRCh38
rs1054988203 Chr.5: 138181338-138181338 on Build GRCh38
rs1039071923 Chr.5: 138181413-138181413 on Build GRCh38
rs376212126 Chr.5: 138181501-138181501 on Build GRCh38
rs779570677 Chr.5: 138181381-138181381 on Build GRCh38
rs769654647 Chr.5: 138181409-138181409 on Build GRCh38
rs747159808 Chr.5: 138181514-138181514 on Build GRCh38
rs781374555 Chr.5: 138181518-138181518 on Build GRCh38
rs76413256 Chr.5: 138181488-138181488 on Build GRCh38

View Full Product Details