Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135895660-135895887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL9
Entrez Gene ID
3578
Gene Name
interleukin 9
Gene Aliases
HP40, IL-9, P40
UniGene
Hs.960
Gene Chromosome Location
Chr.5: 135892246-135895827 on Build GRCh38
Transcripts
NM_000590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4397349 c.44C>T p.S15F Chr.5: 135895773-135895773 on Build GRCh38
5717268 c.22A>G p.T8A Chr.5: 135895795-135895795 on Build GRCh38
3975169 c.75G>A p.A25A Chr.5: 135895742-135895742 on Build GRCh38
735636 c.78G>A p.G26G Chr.5: 135895739-135895739 on Build GRCh38
6169148 c.36C>T p.L12L Chr.5: 135895781-135895781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs982697871 Chr.5: 135895832-135895832 on Build GRCh38
rs772895208 Chr.5: 135895773-135895773 on Build GRCh38
rs753141624 Chr.5: 135895815-135895815 on Build GRCh38
rs749311227 Chr.5: 135895765-135895765 on Build GRCh38
rs755648595 Chr.5: 135895688-135895688 on Build GRCh38
rs536770706 Chr.5: 135895825-135895825 on Build GRCh38
rs774133953 Chr.5: 135895771-135895771 on Build GRCh38
rs758874456 Chr.5: 135895816-135895816 on Build GRCh38
rs908494060 Chr.5: 135895835-135895835 on Build GRCh38
rs751912032 Chr.5: 135895837-135895837 on Build GRCh38
rs770041351 Chr.5: 135895757-135895757 on Build GRCh38
rs781542139 Chr.5: 135895858-135895858 on Build GRCh38
rs558577259 Chr.5: 135895859-135895859 on Build GRCh38
rs530252896 Chr.5: 135895838-135895838 on Build GRCh38
rs768479610 Chr.5: 135895770-135895770 on Build GRCh38
rs868627777 Chr.5: 135895780-135895780 on Build GRCh38
rs201454539 Chr.5: 135895693-135895693 on Build GRCh38
rs758137112 Chr.5: 135895703-135895716 on Build GRCh38
rs765973842 Chr.5: 135895812-135895812 on Build GRCh38
rs777714256 Chr.5: 135895755-135895755 on Build GRCh38
rs368804746 Chr.5: 135895742-135895742 on Build GRCh38
rs765698493 Chr.5: 135895689-135895689 on Build GRCh38
rs747339286 Chr.5: 135895740-135895740 on Build GRCh38
rs758674734 Chr.5: 135895727-135895727 on Build GRCh38
rs760158998 Chr.5: 135895776-135895776 on Build GRCh38
rs949073096 Chr.5: 135895692-135895692 on Build GRCh38
rs975248428 Chr.5: 135895754-135895754 on Build GRCh38
rs923834612 Chr.5: 135895749-135895749 on Build GRCh38
rs936508583 Chr.5: 135895799-135895799 on Build GRCh38
rs775274460 Chr.5: 135895759-135895759 on Build GRCh38
rs964156835 Chr.5: 135895720-135895720 on Build GRCh38
rs569600530 Chr.5: 135895772-135895772 on Build GRCh38
rs778265553 Chr.5: 135895728-135895728 on Build GRCh38
rs749883097 Chr.5: 135895683-135895683 on Build GRCh38
rs373713337 Chr.5: 135895700-135895700 on Build GRCh38
rs748036907 Chr.5: 135895750-135895750 on Build GRCh38

View Full Product Details