Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 85053003-85053266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCMF1
Entrez Gene ID
56888
Gene Name
potassium channel modulatory factor 1
Gene Aliases
DEBT91, FIGC, PCMF, ZZZ1
UniGene
Hs.654968
Gene Chromosome Location
Chr.2: 84971108-85059472 on Build GRCh38
Transcripts
NM_020122.4 XM_006712052.3 XM_011532990.1 XM_017004511.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6690404 c.938G>A p.R313H Chr.2: 85053201-85053201 on Build GRCh38
1185952 c.911C>T p.T304M Chr.2: 85053174-85053174 on Build GRCh38
111462 c.1002delA p.S335fs*19 Chr.2: 85053265-85053265 on Build GRCh38
3001519 c.1001C>A p.S334* Chr.2: 85053264-85053264 on Build GRCh38
4391578 c.893A>C p.D298A Chr.2: 85053156-85053156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374601099 Chr.2: 85053174-85053174 on Build GRCh38
rs764542476 Chr.2: 85053149-85053149 on Build GRCh38
rs757961496 Chr.2: 85053141-85053142 on Build GRCh38
rs748924690 Chr.2: 85053201-85053201 on Build GRCh38
rs998372830 Chr.2: 85053093-85053093 on Build GRCh38
rs779580766 Chr.2: 85053200-85053200 on Build GRCh38
rs752041246 Chr.2: 85053165-85053165 on Build GRCh38
rs567207508 Chr.2: 85053034-85053034 on Build GRCh38
rs746409862 Chr.2: 85053179-85053179 on Build GRCh38
rs770313908 Chr.2: 85053072-85053072 on Build GRCh38
rs768361793 Chr.2: 85053210-85053210 on Build GRCh38
rs1022723853 Chr.2: 85053069-85053069 on Build GRCh38
rs755541595 Chr.2: 85053184-85053184 on Build GRCh38
rs201043553 Chr.2: 85053128-85053128 on Build GRCh38
rs749770581 Chr.2: 85053189-85053189 on Build GRCh38
rs781494825 Chr.2: 85053175-85053175 on Build GRCh38
rs200520798 Chr.2: 85053119-85053119 on Build GRCh38
rs902236433 Chr.2: 85053071-85053071 on Build GRCh38
rs770405567 Chr.2: 85053209-85053209 on Build GRCh38

View Full Product Details