Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 35448098-35448371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0319L
Entrez Gene ID
79932
Gene Name
KIAA0319 like
Gene Aliases
AAVR
UniGene
Hs.456507
Gene Chromosome Location
Chr.1: 35433490-35557670 on Build GRCh38
Transcripts
NM_024874.4 XM_006710907.1 XM_006710909.1 XM_006710910.1 XM_011542179.2 XM_011542180.1 XM_017002366.1 XM_017002367.1 XM_017002368.1 XM_017002369.1 XM_017002370.1 XM_017002371.1 XM_017002372.1 XM_017002373.1 XM_017002374.1 XM_017002375.1 XM_017002376.1 XM_017002377.1 XM_017002378.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7135241 c.2385C>G p.I795M Chr.1: 35448301-35448301 on Build GRCh38
5794480 c.2493T>G p.I831M Chr.1: 35448193-35448193 on Build GRCh38
7112078 c.2494C>A p.Q832K Chr.1: 35448192-35448192 on Build GRCh38
5977954 c.2363A>G p.K788R Chr.1: 35448323-35448323 on Build GRCh38
2080408 c.2454C>T p.L818L Chr.1: 35448232-35448232 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148223068 Chr.1: 35448244-35448244 on Build GRCh38
rs554050475 Chr.1: 35448155-35448155 on Build GRCh38
rs533866704 Chr.1: 35448134-35448134 on Build GRCh38
rs562969254 Chr.1: 35448135-35448135 on Build GRCh38
rs750477600 Chr.1: 35448316-35448316 on Build GRCh38
rs142124223 Chr.1: 35448323-35448323 on Build GRCh38
rs147814828 Chr.1: 35448178-35448178 on Build GRCh38
rs866437712 Chr.1: 35448270-35448270 on Build GRCh38
rs751315305 Chr.1: 35448154-35448154 on Build GRCh38
rs774329290 Chr.1: 35448248-35448248 on Build GRCh38
rs759328053 Chr.1: 35448181-35448181 on Build GRCh38
rs757439687 Chr.1: 35448290-35448290 on Build GRCh38
rs1008172932 Chr.1: 35448278-35448278 on Build GRCh38
rs755365624 Chr.1: 35448343-35448343 on Build GRCh38
rs766149995 Chr.1: 35448136-35448136 on Build GRCh38
rs12729157 Chr.1: 35448251-35448251 on Build GRCh38
rs1000669136 Chr.1: 35448195-35448195 on Build GRCh38
rs747570768 Chr.1: 35448326-35448326 on Build GRCh38
rs758574149 Chr.1: 35448322-35448322 on Build GRCh38
rs1361040 Chr.1: 35448175-35448175 on Build GRCh38
rs767339061 Chr.1: 35448260-35448260 on Build GRCh38
rs150251578 Chr.1: 35448288-35448288 on Build GRCh38
rs200635190 Chr.1: 35448207-35448207 on Build GRCh38
rs769309121 Chr.1: 35448237-35448237 on Build GRCh38
rs867312012 Chr.1: 35448254-35448254 on Build GRCh38
rs778888195 Chr.1: 35448182-35448182 on Build GRCh38
rs759302592 Chr.1: 35448250-35448250 on Build GRCh38
rs956608216 Chr.1: 35448249-35448249 on Build GRCh38
rs776404576 Chr.1: 35448213-35448213 on Build GRCh38
rs775505999 Chr.1: 35448214-35448214 on Build GRCh38
rs765300094 Chr.1: 35448300-35448300 on Build GRCh38
rs965106962 Chr.1: 35448204-35448204 on Build GRCh38
rs147017356 Chr.1: 35448187-35448187 on Build GRCh38
rs764704438 Chr.1: 35448125-35448125 on Build GRCh38
rs761397561 Chr.1: 35448124-35448124 on Build GRCh38
rs963681462 Chr.1: 35448307-35448307 on Build GRCh38
rs1012216372 Chr.1: 35448161-35448161 on Build GRCh38
rs773884840 Chr.1: 35448220-35448222 on Build GRCh38
rs375205434 Chr.1: 35448289-35448289 on Build GRCh38
rs142007809 Chr.1: 35448188-35448188 on Build GRCh38
rs867138388 Chr.1: 35448234-35448234 on Build GRCh38
rs780402859 Chr.1: 35448191-35448191 on Build GRCh38

View Full Product Details