Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 142383695-142383962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRG6
Entrez Gene ID
57211
Gene Name
adhesion G protein-coupled receptor G6
Gene Aliases
APG1, DREG, GPR126, LCCS9, PS1TP2, VIGR
UniGene
Hs.743302
Gene Chromosome Location
Chr.6: 142301919-142446266 on Build GRCh38
Transcripts
NM_001032394.2 NM_001032395.2 NM_020455.5 NM_198569.2 XM_005267061.3 XM_006715516.2 XM_006715517.2 XM_006715518.2 XM_011535964.1 XM_017011085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
123421 c.1153C>G p.P385A Chr.6: 142383774-142383774 on Build GRCh38
4768412 c.1206C>A p.I402I Chr.6: 142383827-142383827 on Build GRCh38
177617 c.1217G>T p.R406M Chr.6: 142383838-142383838 on Build GRCh38
1073960 c.1206C>T p.I402I Chr.6: 142383827-142383827 on Build GRCh38
7142690 c.1207G>T p.D403Y Chr.6: 142383828-142383828 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370831198 Chr.6: 142383828-142383828 on Build GRCh38
rs370523517 Chr.6: 142383737-142383737 on Build GRCh38
rs567380388 Chr.6: 142383897-142383897 on Build GRCh38
rs767749150 Chr.6: 142383911-142383911 on Build GRCh38
rs745369352 Chr.6: 142383889-142383889 on Build GRCh38
rs375001083 Chr.6: 142383834-142383834 on Build GRCh38
rs990925864 Chr.6: 142383922-142383922 on Build GRCh38
rs755936690 Chr.6: 142383854-142383854 on Build GRCh38
rs752532047 Chr.6: 142383747-142383747 on Build GRCh38
rs764376215 Chr.6: 142383723-142383724 on Build GRCh38
rs939267504 Chr.6: 142383917-142383917 on Build GRCh38
rs1030676761 Chr.6: 142383739-142383739 on Build GRCh38
rs182651892 Chr.6: 142383810-142383810 on Build GRCh38
rs777039172 Chr.6: 142383800-142383800 on Build GRCh38
rs76549509 Chr.6: 142383884-142383884 on Build GRCh38
rs755885367 Chr.6: 142383748-142383748 on Build GRCh38
rs964734929 Chr.6: 142383722-142383722 on Build GRCh38
rs753485039 Chr.6: 142383877-142383877 on Build GRCh38
rs762107477 Chr.6: 142383801-142383801 on Build GRCh38
rs752548462 Chr.6: 142383848-142383848 on Build GRCh38
rs756904199 Chr.6: 142383879-142383879 on Build GRCh38
rs919985409 Chr.6: 142383890-142383890 on Build GRCh38
rs769148965 Chr.6: 142383785-142383785 on Build GRCh38
rs770300960 Chr.6: 142383774-142383774 on Build GRCh38
rs538142543 Chr.6: 142383902-142383902 on Build GRCh38
rs1055157924 Chr.6: 142383909-142383909 on Build GRCh38
rs201512134 Chr.6: 142383756-142383756 on Build GRCh38
rs781580053 Chr.6: 142383873-142383873 on Build GRCh38
rs191370055 Chr.6: 142383923-142383923 on Build GRCh38
rs778465876 Chr.6: 142383782-142383782 on Build GRCh38
rs139866989 Chr.6: 142383858-142383858 on Build GRCh38
rs989522828 Chr.6: 142383878-142383878 on Build GRCh38
rs907754398 Chr.6: 142383830-142383830 on Build GRCh38
rs764286860 Chr.6: 142383783-142383783 on Build GRCh38
rs368646880 Chr.6: 142383827-142383827 on Build GRCh38
rs41289839 Chr.6: 142383933-142383933 on Build GRCh38
rs767544205 Chr.6: 142383841-142383841 on Build GRCh38
rs757488548 Chr.6: 142383896-142383896 on Build GRCh38
rs770007435 Chr.6: 142383805-142383805 on Build GRCh38
rs759609818 Chr.6: 142383840-142383840 on Build GRCh38
rs186581090 Chr.6: 142383915-142383915 on Build GRCh38
rs748864894 Chr.6: 142383772-142383772 on Build GRCh38
rs1016273399 Chr.6: 142383798-142383799 on Build GRCh38
rs998618793 Chr.6: 142383815-142383815 on Build GRCh38

View Full Product Details