Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184054670-184054941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTR3C
Entrez Gene ID
170572
Gene Name
5-hydroxytryptamine receptor 3C
Gene Aliases
-
UniGene
Hs.632579
Gene Chromosome Location
Chr.3: 184053047-184060673 on Build GRCh38
Transcripts
NM_130770.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4186069 c.227T>G p.L76R Chr.3: 184054880-184054880 on Build GRCh38
4841218 c.85A>C p.T29P Chr.3: 184054738-184054738 on Build GRCh38
7153160 c.90C>G p.I30M Chr.3: 184054743-184054743 on Build GRCh38
4929132 c.180C>A p.Y60* Chr.3: 184054833-184054833 on Build GRCh38
363371 c.193C>A p.R65S Chr.3: 184054846-184054846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771861231 Chr.3: 184054816-184054816 on Build GRCh38
rs748587586 Chr.3: 184054711-184054711 on Build GRCh38
rs376521137 Chr.3: 184054918-184054918 on Build GRCh38
rs960008234 Chr.3: 184054840-184054840 on Build GRCh38
rs915827411 Chr.3: 184054788-184054788 on Build GRCh38
rs545664364 Chr.3: 184054815-184054815 on Build GRCh38
rs145827675 Chr.3: 184054905-184054905 on Build GRCh38
rs925250793 Chr.3: 184054899-184054899 on Build GRCh38
rs369321709 Chr.3: 184054817-184054817 on Build GRCh38
rs773852375 Chr.3: 184054728-184054728 on Build GRCh38
rs200230893 Chr.3: 184054795-184054795 on Build GRCh38
rs180785188 Chr.3: 184054833-184054833 on Build GRCh38
rs904284678 Chr.3: 184054906-184054906 on Build GRCh38
rs183919204 Chr.3: 184054847-184054847 on Build GRCh38
rs535157974 Chr.3: 184054879-184054880 on Build GRCh38
rs138478768 Chr.3: 184054737-184054737 on Build GRCh38
rs140690227 Chr.3: 184054730-184054730 on Build GRCh38
rs202226852 Chr.3: 184054858-184054858 on Build GRCh38
rs766506234 Chr.3: 184054741-184054741 on Build GRCh38
rs748741355 Chr.3: 184054718-184054718 on Build GRCh38
rs142130329 Chr.3: 184054898-184054898 on Build GRCh38
rs761845667 Chr.3: 184054843-184054843 on Build GRCh38
rs202223050 Chr.3: 184054904-184054904 on Build GRCh38
rs60179621 Chr.3: 184054726-184054726 on Build GRCh38
rs148336193 Chr.3: 184054829-184054829 on Build GRCh38
rs754258738 Chr.3: 184054878-184054878 on Build GRCh38
rs947845175 Chr.3: 184054692-184054692 on Build GRCh38
rs768698957 Chr.3: 184054732-184054732 on Build GRCh38
rs143289420 Chr.3: 184054731-184054731 on Build GRCh38
rs761149070 Chr.3: 184054846-184054846 on Build GRCh38
rs972118774 Chr.3: 184054820-184054820 on Build GRCh38
rs757783719 Chr.3: 184054879-184054879 on Build GRCh38
rs747798264 Chr.3: 184054746-184054746 on Build GRCh38
rs373746183 Chr.3: 184054745-184054745 on Build GRCh38
rs146110065 Chr.3: 184054762-184054762 on Build GRCh38
rs745557511 Chr.3: 184054729-184054729 on Build GRCh38
rs767723576 Chr.3: 184054862-184054862 on Build GRCh38
rs915956601 Chr.3: 184054855-184054855 on Build GRCh38
rs377273716 Chr.3: 184054700-184054700 on Build GRCh38
rs369506821 Chr.3: 184054713-184054713 on Build GRCh38
rs376500343 Chr.3: 184054752-184054752 on Build GRCh38
rs761003444 Chr.3: 184054867-184054867 on Build GRCh38
rs372897520 Chr.3: 184054877-184054877 on Build GRCh38
rs143066269 Chr.3: 184054754-184054754 on Build GRCh38
rs201512118 Chr.3: 184054861-184054861 on Build GRCh38
rs778311364 Chr.3: 184054717-184054717 on Build GRCh38
rs756672085 Chr.3: 184054766-184054766 on Build GRCh38
rs973612301 Chr.3: 184054811-184054811 on Build GRCh38
rs781571969 Chr.3: 184054701-184054701 on Build GRCh38
rs556123858 Chr.3: 184054761-184054761 on Build GRCh38
rs550609003 Chr.3: 184054865-184054865 on Build GRCh38
rs375625828 Chr.3: 184054892-184054892 on Build GRCh38
rs754415640 Chr.3: 184054764-184054764 on Build GRCh38

View Full Product Details