Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32158715-32158962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPD1L
Entrez Gene ID
23171
Gene Name
glycerol-3-phosphate dehydrogenase 1-like
Gene Aliases
GPD1-L
UniGene
Hs.82432
Gene Chromosome Location
Chr.3: 32106511-32168715 on Build GRCh38
Transcripts
NM_015141.3 XM_006713068.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
252529 c.691C>T p.R231C Chr.3: 32158948-32158948 on Build GRCh38
5823598 c.624C>T p.I208I Chr.3: 32158881-32158881 on Build GRCh38
5174267 c.624C>A p.I208I Chr.3: 32158881-32158881 on Build GRCh38
7348090 c.652G>A p.G218S Chr.3: 32158909-32158909 on Build GRCh38
5059648 c.682G>A p.A228T Chr.3: 32158939-32158939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748260612 Chr.3: 32158835-32158835 on Build GRCh38
rs752064476 Chr.3: 32158829-32158829 on Build GRCh38
rs557211491 Chr.3: 32158787-32158787 on Build GRCh38
rs769819874 Chr.3: 32158842-32158842 on Build GRCh38
rs72546648 Chr.3: 32158916-32158916 on Build GRCh38
rs780462232 Chr.3: 32158828-32158828 on Build GRCh38
rs749527173 Chr.3: 32158936-32158936 on Build GRCh38
rs534799892 Chr.3: 32158848-32158848 on Build GRCh38
rs965373652 Chr.3: 32158863-32158863 on Build GRCh38
rs891624963 Chr.3: 32158812-32158812 on Build GRCh38
rs755297625 Chr.3: 32158906-32158906 on Build GRCh38
rs267599768 Chr.3: 32158881-32158881 on Build GRCh38
rs935075011 Chr.3: 32158767-32158767 on Build GRCh38
rs559530222 Chr.3: 32158909-32158909 on Build GRCh38
rs980532320 Chr.3: 32158937-32158937 on Build GRCh38
rs758710275 Chr.3: 32158845-32158845 on Build GRCh38
rs145648143 Chr.3: 32158905-32158905 on Build GRCh38
rs373325536 Chr.3: 32158887-32158887 on Build GRCh38
rs911671433 Chr.3: 32158758-32158758 on Build GRCh38
rs1010779006 Chr.3: 32158825-32158825 on Build GRCh38
rs1051418203 Chr.3: 32158757-32158757 on Build GRCh38
rs760328411 Chr.3: 32158864-32158864 on Build GRCh38
rs148361833 Chr.3: 32158910-32158910 on Build GRCh38
rs1057393467 Chr.3: 32158807-32158807 on Build GRCh38
rs2305361 Chr.3: 32158830-32158830 on Build GRCh38
rs765893811 Chr.3: 32158896-32158896 on Build GRCh38
rs775341363 Chr.3: 32158862-32158862 on Build GRCh38
rs545047908 Chr.3: 32158941-32158941 on Build GRCh38
rs2305362 Chr.3: 32158741-32158741 on Build GRCh38
rs750110247 Chr.3: 32158888-32158888 on Build GRCh38
rs1052314006 Chr.3: 32158797-32158797 on Build GRCh38
rs72546647 Chr.3: 32158915-32158915 on Build GRCh38
rs762654723 Chr.3: 32158895-32158895 on Build GRCh38
rs973664942 Chr.3: 32158938-32158938 on Build GRCh38
rs960188961 Chr.3: 32158879-32158879 on Build GRCh38
rs2044880 Chr.3: 32158867-32158867 on Build GRCh38
rs749491704 Chr.3: 32158846-32158846 on Build GRCh38
rs757614405 Chr.3: 32158942-32158942 on Build GRCh38
rs149641866 Chr.3: 32158908-32158908 on Build GRCh38
rs538696684 Chr.3: 32158772-32158772 on Build GRCh38
rs371424363 Chr.3: 32158831-32158831 on Build GRCh38
rs765435378 Chr.3: 32158744-32158744 on Build GRCh38

View Full Product Details