Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 119441768-119442041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK5
Entrez Gene ID
2869
Gene Name
G protein-coupled receptor kinase 5
Gene Aliases
GPRK5
UniGene
Hs.524625
Gene Chromosome Location
Chr.10: 119207685-119455619 on Build GRCh38
Transcripts
NM_005308.2 XM_005269707.1 XM_005269708.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
915353 c.1008C>T p.I336I Chr.10: 119442039-119442039 on Build GRCh38
7297289 c.1010C>T p.P337L Chr.10: 119442041-119442041 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs982821945 Chr.10: 119441949-119441949 on Build GRCh38
rs753350829 Chr.10: 119441966-119441966 on Build GRCh38
rs748682304 Chr.10: 119441990-119441990 on Build GRCh38
rs777370203 Chr.10: 119441797-119441797 on Build GRCh38
rs147251247 Chr.10: 119441920-119441920 on Build GRCh38
rs1020918710 Chr.10: 119441809-119441809 on Build GRCh38
rs767329679 Chr.10: 119441948-119441948 on Build GRCh38
rs952783052 Chr.10: 119441946-119441946 on Build GRCh38
rs144057766 Chr.10: 119441907-119441907 on Build GRCh38
rs952269990 Chr.10: 119441868-119441868 on Build GRCh38
rs1040340443 Chr.10: 119441984-119441984 on Build GRCh38
rs967667264 Chr.10: 119441863-119441863 on Build GRCh38
rs779499424 Chr.10: 119441985-119441985 on Build GRCh38
rs117983291 Chr.10: 119441915-119441915 on Build GRCh38
rs755761601 Chr.10: 119441958-119441958 on Build GRCh38
rs981370320 Chr.10: 119441869-119441870 on Build GRCh38
rs1003105418 Chr.10: 119441873-119441873 on Build GRCh38
rs779623037 Chr.10: 119441965-119441965 on Build GRCh38
rs529863380 Chr.10: 119441805-119441805 on Build GRCh38
rs778166997 Chr.10: 119442006-119442006 on Build GRCh38
rs757774698 Chr.10: 119441896-119441896 on Build GRCh38
rs370757498 Chr.10: 119441974-119441974 on Build GRCh38
rs750255115 Chr.10: 119441952-119441952 on Build GRCh38
rs920853070 Chr.10: 119441839-119441839 on Build GRCh38
rs772515562 Chr.10: 119441993-119441993 on Build GRCh38
rs746552498 Chr.10: 119441803-119441803 on Build GRCh38
rs940605304 Chr.10: 119441975-119441975 on Build GRCh38
rs549590040 Chr.10: 119441818-119441818 on Build GRCh38

View Full Product Details