Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 39346361-39346632 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF6
Entrez Gene ID
221458
Gene Name
kinesin family member 6
Gene Aliases
C6orf102, dJ1043E3.1, dJ137F1.4, dJ188D3.1
UniGene
Hs.588202
Gene Chromosome Location
Chr.6: 39335100-39725405 on Build GRCh38
Transcripts
NM_001289020.1 NM_001289021.1 NM_001289024.1 NM_145027.4 XM_005248904.4 XM_011514357.2 XM_011514358.2 XM_011514359.2 XM_017010427.1 XM_017010428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6329064 c.2211C>T p.G737G Chr.6: 39346496-39346496 on Build GRCh38
232632 c.2194G>A p.E732K Chr.6: 39346513-39346513 on Build GRCh38
5405250 c.552C>T p.V184V Chr.6: 39346508-39346508 on Build GRCh38
4535456 c.2026G>A p.E676K Chr.6: 39346513-39346513 on Build GRCh38
4535457 c.547G>A p.E183K Chr.6: 39346513-39346513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766695790 Chr.6: 39346510-39346510 on Build GRCh38
rs969416959 Chr.6: 39346529-39346529 on Build GRCh38
rs534806340 Chr.6: 39346433-39346433 on Build GRCh38
rs367591537 Chr.6: 39346458-39346458 on Build GRCh38
rs761081302 Chr.6: 39346507-39346507 on Build GRCh38
rs758567725 Chr.6: 39346405-39346405 on Build GRCh38
rs754366412 Chr.6: 39346523-39346523 on Build GRCh38
rs35218076 Chr.6: 39346466-39346466 on Build GRCh38
rs79494296 Chr.6: 39346435-39346435 on Build GRCh38
rs972775803 Chr.6: 39346569-39346569 on Build GRCh38
rs916015197 Chr.6: 39346415-39346415 on Build GRCh38
rs532132719 Chr.6: 39346483-39346483 on Build GRCh38
rs576254527 Chr.6: 39346513-39346513 on Build GRCh38
rs771168241 Chr.6: 39346542-39346542 on Build GRCh38
rs556461887 Chr.6: 39346484-39346484 on Build GRCh38
rs370305271 Chr.6: 39346427-39346427 on Build GRCh38
rs745829292 Chr.6: 39346441-39346441 on Build GRCh38
rs542439920 Chr.6: 39346468-39346468 on Build GRCh38
rs959790590 Chr.6: 39346568-39346568 on Build GRCh38
rs948812859 Chr.6: 39346432-39346432 on Build GRCh38
rs748348071 Chr.6: 39346401-39346401 on Build GRCh38
rs778002387 Chr.6: 39346412-39346412 on Build GRCh38
rs776761240 Chr.6: 39346544-39346544 on Build GRCh38
rs999255199 Chr.6: 39346453-39346453 on Build GRCh38
rs764761617 Chr.6: 39346541-39346541 on Build GRCh38
rs748048932 Chr.6: 39346436-39346436 on Build GRCh38
rs752707041 Chr.6: 39346555-39346555 on Build GRCh38
rs777641240 Chr.6: 39346459-39346459 on Build GRCh38
rs769339529 Chr.6: 39346452-39346452 on Build GRCh38

View Full Product Details