Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54029606-54029875 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXC4
Entrez Gene ID
3221
Gene Name
homeobox C4
Gene Aliases
HOX3, HOX3E, cp19
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54056030 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Gene Symbol
HOXC5
Entrez Gene ID
3222
Gene Name
homeobox C5
Gene Aliases
CP11, HOX3, HOX3D
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54035361 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Gene Symbol
HOXC6
Entrez Gene ID
3223
Gene Name
homeobox C6
Gene Aliases
CP25, HHO.C8, HOX3, HOX3C
UniGene
Hs.549040
Gene Chromosome Location
Chr.12: 54016852-54030823 on Build GRCh38
Transcripts
NM_014620.5 NR_003084.2 NM_004503.3 NM_153693.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6681765 c.417G>A p.A139A Chr.12: 54029671-54029671 on Build GRCh38
1239088 c.457A>G p.T153A Chr.12: 54029711-54029711 on Build GRCh38
416167 c.522C>T p.I174I Chr.12: 54029776-54029776 on Build GRCh38
1188514 c.575G>A p.R192H Chr.12: 54029829-54029829 on Build GRCh38
4043150 c.531G>A p.A177A Chr.12: 54029785-54029785 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976125070 Chr.12: 54029828-54029828 on Build GRCh38
rs151083984 Chr.12: 54029779-54029779 on Build GRCh38
rs756503013 Chr.12: 54029677-54029677 on Build GRCh38
rs1003887304 Chr.12: 54029637-54029637 on Build GRCh38
rs139697680 Chr.12: 54029792-54029792 on Build GRCh38
rs372813456 Chr.12: 54029739-54029739 on Build GRCh38
rs759420807 Chr.12: 54029756-54029756 on Build GRCh38
rs529857999 Chr.12: 54029704-54029704 on Build GRCh38
rs759457160 Chr.12: 54029651-54029651 on Build GRCh38
rs1003317155 Chr.12: 54029700-54029700 on Build GRCh38
rs761472061 Chr.12: 54029781-54029781 on Build GRCh38
rs751242313 Chr.12: 54029676-54029676 on Build GRCh38
rs552181831 Chr.12: 54029797-54029797 on Build GRCh38
rs138685629 Chr.12: 54029675-54029675 on Build GRCh38
rs377095092 Chr.12: 54029730-54029730 on Build GRCh38
rs778998070 Chr.12: 54029695-54029695 on Build GRCh38
rs958915325 Chr.12: 54029722-54029722 on Build GRCh38
rs775505948 Chr.12: 54029833-54029833 on Build GRCh38
rs541354754 Chr.12: 54029759-54029759 on Build GRCh38
rs777768728 Chr.12: 54029840-54029840 on Build GRCh38
rs769891934 Chr.12: 54029757-54029757 on Build GRCh38
rs766189183 Chr.12: 54029642-54029642 on Build GRCh38
rs1036729726 Chr.12: 54029663-54029663 on Build GRCh38
rs762610633 Chr.12: 54029777-54029777 on Build GRCh38
rs548281243 Chr.12: 54029742-54029742 on Build GRCh38
rs754192842 Chr.12: 54029791-54029791 on Build GRCh38
rs773395635 Chr.12: 54029627-54029627 on Build GRCh38
rs977386749 Chr.12: 54029794-54029794 on Build GRCh38
rs375750909 Chr.12: 54029755-54029755 on Build GRCh38
rs565754932 Chr.12: 54029679-54029679 on Build GRCh38
rs113381644 Chr.12: 54029703-54029703 on Build GRCh38
rs752213049 Chr.12: 54029661-54029661 on Build GRCh38
rs770642032 Chr.12: 54029746-54029746 on Build GRCh38
rs776432961 Chr.12: 54029644-54029644 on Build GRCh38
rs763888269 Chr.12: 54029666-54029666 on Build GRCh38
rs753193612 Chr.12: 54029801-54029801 on Build GRCh38
rs199717526 Chr.12: 54029785-54029785 on Build GRCh38
rs760440415 Chr.12: 54029631-54029631 on Build GRCh38
rs751262560 Chr.12: 54029780-54029780 on Build GRCh38
rs755466327 Chr.12: 54029694-54029694 on Build GRCh38
rs764671940 Chr.12: 54029652-54029652 on Build GRCh38
rs773485388 Chr.12: 54029707-54029707 on Build GRCh38
rs761693320 Chr.12: 54029842-54029843 on Build GRCh38
rs775544979 Chr.12: 54029763-54029763 on Build GRCh38
rs61736613 Chr.12: 54029782-54029782 on Build GRCh38
rs749774424 Chr.12: 54029693-54029693 on Build GRCh38
rs757948540 Chr.12: 54029665-54029665 on Build GRCh38
rs143618403 Chr.12: 54029809-54029809 on Build GRCh38
rs368405993 Chr.12: 54029737-54029737 on Build GRCh38

View Full Product Details