Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49038205-49038476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGF2BP1
Entrez Gene ID
10642
Gene Name
insulin like growth factor 2 mRNA binding protein 1
Gene Aliases
CRD-BP, CRDBP, IMP-1, IMP1, VICKZ1, ZBP1
UniGene
Hs.144936
Gene Chromosome Location
Chr.17: 48997412-49056145 on Build GRCh38
Transcripts
NM_001160423.1 NM_006546.3 XM_005256955.3 XM_011524201.2 XM_017024022.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4501420 c.588C>T p.I196I Chr.17: 49038354-49038354 on Build GRCh38
980702 c.596G>A p.R199Q Chr.17: 49038362-49038362 on Build GRCh38
6684234 c.490G>C p.E164Q Chr.17: 49038256-49038256 on Build GRCh38
2154294 c.499C>T p.R167C Chr.17: 49038265-49038265 on Build GRCh38
980701 c.592C>A p.L198I Chr.17: 49038358-49038358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377013730 Chr.17: 49038330-49038330 on Build GRCh38
rs554770601 Chr.17: 49038284-49038284 on Build GRCh38
rs779507141 Chr.17: 49038286-49038286 on Build GRCh38
rs746549698 Chr.17: 49038287-49038287 on Build GRCh38
rs761394589 Chr.17: 49038449-49038449 on Build GRCh38
rs767875851 Chr.17: 49038263-49038263 on Build GRCh38
rs774524369 Chr.17: 49038321-49038321 on Build GRCh38
rs373702055 Chr.17: 49038264-49038264 on Build GRCh38
rs752960224 Chr.17: 49038362-49038362 on Build GRCh38
rs767783216 Chr.17: 49038336-49038336 on Build GRCh38
rs747817650 Chr.17: 49038441-49038441 on Build GRCh38
rs766547480 Chr.17: 49038242-49038242 on Build GRCh38
rs557111796 Chr.17: 49038266-49038266 on Build GRCh38
rs987445536 Chr.17: 49038406-49038406 on Build GRCh38
rs762860271 Chr.17: 49038238-49038238 on Build GRCh38
rs139671755 Chr.17: 49038231-49038231 on Build GRCh38
rs760862684 Chr.17: 49038375-49038375 on Build GRCh38
rs754194926 Chr.17: 49038421-49038421 on Build GRCh38
rs999692017 Chr.17: 49038290-49038290 on Build GRCh38
rs757720909 Chr.17: 49038424-49038424 on Build GRCh38
rs769621595 Chr.17: 49038310-49038310 on Build GRCh38
rs546002680 Chr.17: 49038317-49038317 on Build GRCh38
rs747828911 Chr.17: 49038303-49038303 on Build GRCh38
rs764495682 Chr.17: 49038399-49038399 on Build GRCh38
rs267604935 Chr.17: 49038273-49038273 on Build GRCh38
rs962810679 Chr.17: 49038408-49038408 on Build GRCh38
rs751732919 Chr.17: 49038245-49038245 on Build GRCh38
rs542203174 Chr.17: 49038429-49038429 on Build GRCh38
rs1024617698 Chr.17: 49038269-49038269 on Build GRCh38
rs145184529 Chr.17: 49038432-49038432 on Build GRCh38
rs576996590 Chr.17: 49038289-49038289 on Build GRCh38
rs368043361 Chr.17: 49038298-49038298 on Build GRCh38
rs373305132 Chr.17: 49038320-49038320 on Build GRCh38
rs543797797 Chr.17: 49038249-49038249 on Build GRCh38
rs754486391 Chr.17: 49038436-49038436 on Build GRCh38
rs780628959 Chr.17: 49038437-49038437 on Build GRCh38
rs754354262 Chr.17: 49038274-49038274 on Build GRCh38
rs756506419 Chr.17: 49038268-49038268 on Build GRCh38

View Full Product Details