Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 151294233-151294500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR87
Entrez Gene ID
53836
Gene Name
G protein-coupled receptor 87
Gene Aliases
FKSG78, GPR95, KPG_002
UniGene
Hs.591292
Gene Chromosome Location
Chr.3: 151294086-151316848 on Build GRCh38
Transcripts
NM_023915.3 NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1

Gene Symbol
MED12L
Entrez Gene ID
116931
Gene Name
mediator complex subunit 12 like
Gene Aliases
NOPAR, TNRC11L, TRALP, TRALPUSH
UniGene
Hs.744234
Gene Chromosome Location
Chr.3: 151085665-151436677 on Build GRCh38
Transcripts
NM_023915.3 NM_053002.5 XM_006713487.3 XM_011512390.2 XM_011512394.2 XM_011512399.2 XM_017005676.1 XM_017005677.1 XM_017005678.1 XM_017005679.1 XM_017005680.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1693601 c.995G>A p.R332K Chr.3: 151294251-151294251 on Build GRCh38
5811211 c.908C>T p.S303F Chr.3: 151294338-151294338 on Build GRCh38
3783670 c.748C>T p.H250Y Chr.3: 151294498-151294498 on Build GRCh38
5784543 c.959C>G p.S320* Chr.3: 151294287-151294287 on Build GRCh38
3774644 c.928G>C p.D310H Chr.3: 151294318-151294318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755414425 Chr.3: 151294304-151294304 on Build GRCh38
rs779582675 Chr.3: 151294439-151294439 on Build GRCh38
rs1027857953 Chr.3: 151294329-151294329 on Build GRCh38
rs771180891 Chr.3: 151294373-151294373 on Build GRCh38
rs200612301 Chr.3: 151294444-151294444 on Build GRCh38
rs762139683 Chr.3: 151294273-151294273 on Build GRCh38
rs76418398 Chr.3: 151294467-151294467 on Build GRCh38
rs986712158 Chr.3: 151294392-151294392 on Build GRCh38
rs773522593 Chr.3: 151294292-151294292 on Build GRCh38
rs749558443 Chr.3: 151294458-151294458 on Build GRCh38
rs755718258 Chr.3: 151294414-151294419 on Build GRCh38
rs781148826 Chr.3: 151294349-151294349 on Build GRCh38
rs750135413 Chr.3: 151294427-151294427 on Build GRCh38
rs771307776 Chr.3: 151294459-151294459 on Build GRCh38
rs199886869 Chr.3: 151294436-151294436 on Build GRCh38
rs747019751 Chr.3: 151294341-151294341 on Build GRCh38
rs779527569 Chr.3: 151294339-151294339 on Build GRCh38
rs987770351 Chr.3: 151294350-151294350 on Build GRCh38
rs199868921 Chr.3: 151294450-151294450 on Build GRCh38
rs776272233 Chr.3: 151294272-151294272 on Build GRCh38
rs754466461 Chr.3: 151294443-151294443 on Build GRCh38
rs767915012 Chr.3: 151294383-151294383 on Build GRCh38
rs1055230321 Chr.3: 151294445-151294445 on Build GRCh38
rs144965574 Chr.3: 151294358-151294358 on Build GRCh38
rs767931824 Chr.3: 151294401-151294401 on Build GRCh38
rs774582377 Chr.3: 151294460-151294460 on Build GRCh38
rs746916197 Chr.3: 151294268-151294268 on Build GRCh38
rs575711330 Chr.3: 151294380-151294380 on Build GRCh38
rs752166018 Chr.3: 151294302-151294302 on Build GRCh38
rs757972503 Chr.3: 151294338-151294338 on Build GRCh38
rs112156636 Chr.3: 151294396-151294396 on Build GRCh38
rs371872806 Chr.3: 151294475-151294475 on Build GRCh38
rs374834030 Chr.3: 151294477-151294477 on Build GRCh38
rs765411125 Chr.3: 151294287-151294287 on Build GRCh38
rs750011984 Chr.3: 151294336-151294336 on Build GRCh38
rs778550664 Chr.3: 151294335-151294335 on Build GRCh38
rs769641930 Chr.3: 151294367-151294367 on Build GRCh38
rs761166294 Chr.3: 151294421-151294421 on Build GRCh38
rs966287151 Chr.3: 151294411-151294411 on Build GRCh38
rs760952687 Chr.3: 151294410-151294410 on Build GRCh38
rs777117744 Chr.3: 151294416-151294420 on Build GRCh38
rs372384562 Chr.3: 151294370-151294370 on Build GRCh38
rs767964182 Chr.3: 151294322-151294322 on Build GRCh38
rs777686347 Chr.3: 151294455-151294455 on Build GRCh38
rs746334545 Chr.3: 151294296-151294296 on Build GRCh38
rs768554792 Chr.3: 151294270-151294270 on Build GRCh38
rs972749154 Chr.3: 151294385-151294385 on Build GRCh38
rs773631218 Chr.3: 151294369-151294369 on Build GRCh38
rs772734660 Chr.3: 151294357-151294357 on Build GRCh38
rs766560001 Chr.3: 151294301-151294301 on Build GRCh38
rs752278985 Chr.3: 151294401-151294404 on Build GRCh38
rs753035114 Chr.3: 151294325-151294325 on Build GRCh38
rs149904899 Chr.3: 151294467-151294468 on Build GRCh38
rs751063950 Chr.3: 151294441-151294441 on Build GRCh38
rs753189066 Chr.3: 151294405-151294405 on Build GRCh38
rs539111679 Chr.3: 151294334-151294334 on Build GRCh38
rs759547383 Chr.3: 151294465-151294465 on Build GRCh38
rs184212480 Chr.3: 151294346-151294346 on Build GRCh38
rs375414416 Chr.3: 151294375-151294375 on Build GRCh38
rs961734481 Chr.3: 151294285-151294285 on Build GRCh38
rs767203300 Chr.3: 151294338-151294338 on Build GRCh38

View Full Product Details