Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42165360-42165627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNH4
Entrez Gene ID
23415
Gene Name
potassium voltage-gated channel subfamily H member 4
Gene Aliases
BEC2, ELK1, Kv12.3
UniGene
Hs.304081
Gene Chromosome Location
Chr.17: 42156892-42181278 on Build GRCh38
Transcripts
NM_012285.2 XM_017024400.1 XM_017024401.1 XM_017024402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5364363 c.2085+2T>C p.? Chr.17: 42165447-42165447 on Build GRCh38
4824288 c.2044C>G p.R682G Chr.17: 42165490-42165490 on Build GRCh38
4433822 c.1979G>A p.R660Q Chr.17: 42165555-42165555 on Build GRCh38
4532990 c.1938G>A p.V646V Chr.17: 42165596-42165596 on Build GRCh38
5496637 c.1950C>T p.T650T Chr.17: 42165584-42165584 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754820216 Chr.17: 42165427-42165427 on Build GRCh38
rs1032725778 Chr.17: 42165518-42165518 on Build GRCh38
rs922279261 Chr.17: 42165584-42165584 on Build GRCh38
rs773858322 Chr.17: 42165474-42165474 on Build GRCh38
rs953491396 Chr.17: 42165532-42165532 on Build GRCh38
rs781276359 Chr.17: 42165432-42165432 on Build GRCh38
rs149532318 Chr.17: 42165490-42165490 on Build GRCh38
rs956503408 Chr.17: 42165431-42165431 on Build GRCh38
rs747322468 Chr.17: 42165430-42165430 on Build GRCh38
rs761467739 Chr.17: 42165488-42165488 on Build GRCh38
rs754763105 Chr.17: 42165505-42165505 on Build GRCh38
rs562415310 Chr.17: 42165422-42165422 on Build GRCh38
rs1038388702 Chr.17: 42165400-42165400 on Build GRCh38
rs769299655 Chr.17: 42165591-42165591 on Build GRCh38
rs144160163 Chr.17: 42165547-42165547 on Build GRCh38
rs760127990 Chr.17: 42165497-42165497 on Build GRCh38
rs765074595 Chr.17: 42165519-42165519 on Build GRCh38
rs745941682 Chr.17: 42165436-42165436 on Build GRCh38
rs776187068 Chr.17: 42165462-42165462 on Build GRCh38
rs79383250 Chr.17: 42165556-42165556 on Build GRCh38
rs766285729 Chr.17: 42165498-42165498 on Build GRCh38
rs752450441 Chr.17: 42165530-42165530 on Build GRCh38
rs572218290 Chr.17: 42165535-42165535 on Build GRCh38
rs777944981 Chr.17: 42165428-42165428 on Build GRCh38
rs976234402 Chr.17: 42165452-42165452 on Build GRCh38
rs955774151 Chr.17: 42165555-42165555 on Build GRCh38
rs749868044 Chr.17: 42165581-42165581 on Build GRCh38
rs780702177 Chr.17: 42165576-42165576 on Build GRCh38
rs200351279 Chr.17: 42165469-42165469 on Build GRCh38
rs780000357 Chr.17: 42165411-42165411 on Build GRCh38
rs781290348 Chr.17: 42165546-42165546 on Build GRCh38
rs770373624 Chr.17: 42165439-42165439 on Build GRCh38
rs774709267 Chr.17: 42165602-42165602 on Build GRCh38
rs767078879 Chr.17: 42165489-42165489 on Build GRCh38
rs757435873 Chr.17: 42165538-42165538 on Build GRCh38
rs191329438 Chr.17: 42165468-42165468 on Build GRCh38
rs753817153 Chr.17: 42165504-42165504 on Build GRCh38

View Full Product Details