Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48694076-48694325 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IP6K2
Entrez Gene ID
51447
Gene Name
inositol hexakisphosphate kinase 2
Gene Aliases
IHPK2, PIUS
UniGene
Hs.595983
Gene Chromosome Location
Chr.3: 48688003-48717278 on Build GRCh38
Transcripts
NM_001005909.2 NM_001005910.2 NM_001005911.2 NM_001146178.2 NM_001146179.2 NM_001190316.1 NM_001190317.1 NM_016291.3 NR_027437.2 NR_027438.2 XM_006713199.3 XM_006713200.1 XM_006713201.1 XM_006713202.1 XM_011533816.1 XM_011533817.1 XM_011533818.1 XM_017006583.1 XM_017006584.1 XM_017006585.1 XM_017006586.1 XM_017006587.1 XM_017006589.1 XM_017006590.1 XM_017006591.1 XM_017006593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4829976 c.293G>C p.*98S Chr.3: 48694150-48694150 on Build GRCh38
5532772 c.265C>T p.P89S Chr.3: 48694178-48694178 on Build GRCh38
1045950 c.203G>T p.G68V Chr.3: 48694240-48694240 on Build GRCh38
4646715 c.229T>C p.W77R Chr.3: 48694214-48694214 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs952708678 Chr.3: 48694238-48694238 on Build GRCh38
rs1028319701 Chr.3: 48694205-48694205 on Build GRCh38
rs1054004073 Chr.3: 48694127-48694127 on Build GRCh38
rs901489097 Chr.3: 48694244-48694245 on Build GRCh38
rs766471836 Chr.3: 48694160-48694160 on Build GRCh38
rs755526077 Chr.3: 48694172-48694172 on Build GRCh38
rs562135569 Chr.3: 48694289-48694289 on Build GRCh38
rs2077960 Chr.3: 48694102-48694102 on Build GRCh38
rs866280050 Chr.3: 48694183-48694183 on Build GRCh38
rs111512037 Chr.3: 48694169-48694169 on Build GRCh38
rs984445927 Chr.3: 48694252-48694252 on Build GRCh38
rs1043995913 Chr.3: 48694136-48694136 on Build GRCh38
rs1007733621 Chr.3: 48694164-48694164 on Build GRCh38
rs550916337 Chr.3: 48694137-48694137 on Build GRCh38
rs369362908 Chr.3: 48694142-48694142 on Build GRCh38
rs781485112 Chr.3: 48694198-48694198 on Build GRCh38
rs996888660 Chr.3: 48694171-48694171 on Build GRCh38
rs892647108 Chr.3: 48694128-48694128 on Build GRCh38
rs936928402 Chr.3: 48694107-48694107 on Build GRCh38
rs758648194 Chr.3: 48694106-48694106 on Build GRCh38
rs376953443 Chr.3: 48694143-48694143 on Build GRCh38
rs752683798 Chr.3: 48694257-48694257 on Build GRCh38

View Full Product Details