Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206798802-206799057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL19
Entrez Gene ID
29949
Gene Name
interleukin 19
Gene Aliases
IL-10C, MDA1, NG.1, ZMDA1
UniGene
Hs.661017
Gene Chromosome Location
Chr.1: 206798870-206842981 on Build GRCh38
Transcripts
NM_153758.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5413707 c.8C>T p.T3I Chr.1: 206798902-206798902 on Build GRCh38
4479165 c.22C>T p.P8S Chr.1: 206798916-206798916 on Build GRCh38
1689742 c.73C>T p.H25Y Chr.1: 206798967-206798967 on Build GRCh38
5379437 c.99C>T p.G33G Chr.1: 206798993-206798993 on Build GRCh38
1473330 c.6C>A p.C2* Chr.1: 206798900-206798900 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777031399 Chr.1: 206798968-206798968 on Build GRCh38
rs989199735 Chr.1: 206798959-206798959 on Build GRCh38
rs148873337 Chr.1: 206798919-206798919 on Build GRCh38
rs762542493 Chr.1: 206799004-206799004 on Build GRCh38
rs186592422 Chr.1: 206798842-206798842 on Build GRCh38
rs769488868 Chr.1: 206798987-206798987 on Build GRCh38
rs141906021 Chr.1: 206799003-206799003 on Build GRCh38
rs144004857 Chr.1: 206798930-206798930 on Build GRCh38
rs748587989 Chr.1: 206798863-206798863 on Build GRCh38
rs748266118 Chr.1: 206798940-206798940 on Build GRCh38
rs778367541 Chr.1: 206798961-206798961 on Build GRCh38
rs773384170 Chr.1: 206798883-206798883 on Build GRCh38
rs767460276 Chr.1: 206798924-206798924 on Build GRCh38
rs758774648 Chr.1: 206798948-206798948 on Build GRCh38
rs966367868 Chr.1: 206798984-206798984 on Build GRCh38
rs1004175069 Chr.1: 206798886-206798886 on Build GRCh38
rs754089269 Chr.1: 206798929-206798929 on Build GRCh38
rs761042152 Chr.1: 206798885-206798885 on Build GRCh38
rs770522987 Chr.1: 206798899-206798899 on Build GRCh38
rs541491542 Chr.1: 206798912-206798912 on Build GRCh38
rs776300647 Chr.1: 206798903-206798903 on Build GRCh38
rs778855635 Chr.1: 206798938-206798938 on Build GRCh38
rs982157813 Chr.1: 206799002-206799002 on Build GRCh38
rs957819350 Chr.1: 206798943-206798943 on Build GRCh38
rs759229160 Chr.1: 206798910-206798910 on Build GRCh38
rs745509694 Chr.1: 206798971-206798971 on Build GRCh38
rs779097284 Chr.1: 206798862-206798862 on Build GRCh38
rs757149201 Chr.1: 206798921-206798921 on Build GRCh38
rs1056353851 Chr.1: 206798822-206798822 on Build GRCh38
rs568182051 Chr.1: 206799033-206799033 on Build GRCh38
rs964435319 Chr.1: 206799001-206799001 on Build GRCh38
rs76513170 Chr.1: 206798962-206798962 on Build GRCh38
rs761889997 Chr.1: 206799023-206799023 on Build GRCh38
rs116126622 Chr.1: 206798864-206798864 on Build GRCh38
rs780442485 Chr.1: 206798845-206798845 on Build GRCh38
rs894582322 Chr.1: 206798823-206798823 on Build GRCh38
rs374010519 Chr.1: 206799011-206799011 on Build GRCh38
rs143483780 Chr.1: 206798917-206798917 on Build GRCh38
rs749340544 Chr.1: 206798853-206798853 on Build GRCh38
rs201550039 Chr.1: 206798953-206798953 on Build GRCh38
rs764102395 Chr.1: 206798918-206798918 on Build GRCh38
rs532313651 Chr.1: 206798859-206798859 on Build GRCh38

View Full Product Details