Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 55439363-55439622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGCLL1
Entrez Gene ID
54511
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1
Gene Aliases
bA418P12.1, er-cHL
UniGene
Hs.147054
Gene Chromosome Location
Chr.6: 55434373-55579214 on Build GRCh38
Transcripts
NM_001042406.1 NM_001287741.1 NM_001287746.1 NM_001287753.1 NM_019036.2 NR_109869.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1734915 c.916T>G p.S306A Chr.6: 55439529-55439529 on Build GRCh38
5870946 c.908C>T p.S303F Chr.6: 55439537-55439537 on Build GRCh38
1167666 c.947delG p.G316fs*6 Chr.6: 55439498-55439498 on Build GRCh38
5912439 c.973G>A p.D325N Chr.6: 55439472-55439472 on Build GRCh38
6299483 c.887T>A p.M296K Chr.6: 55439558-55439558 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925832427 Chr.6: 55439416-55439416 on Build GRCh38
rs745740684 Chr.6: 55439446-55439446 on Build GRCh38
rs374968414 Chr.6: 55439559-55439559 on Build GRCh38
rs183132034 Chr.6: 55439441-55439441 on Build GRCh38
rs781593132 Chr.6: 55439445-55439446 on Build GRCh38
rs759038429 Chr.6: 55439393-55439393 on Build GRCh38
rs868698183 Chr.6: 55439444-55439445 on Build GRCh38
rs896718116 Chr.6: 55439544-55439544 on Build GRCh38
rs763493743 Chr.6: 55439502-55439502 on Build GRCh38
rs186128280 Chr.6: 55439536-55439536 on Build GRCh38
rs367874288 Chr.6: 55439554-55439554 on Build GRCh38
rs550221893 Chr.6: 55439512-55439512 on Build GRCh38
rs374502890 Chr.6: 55439392-55439392 on Build GRCh38
rs746837826 Chr.6: 55439457-55439457 on Build GRCh38
rs148276847 Chr.6: 55439527-55439527 on Build GRCh38
rs748333297 Chr.6: 55439573-55439573 on Build GRCh38
rs773428478 Chr.6: 55439499-55439499 on Build GRCh38
rs769911043 Chr.6: 55439484-55439484 on Build GRCh38
rs930768439 Chr.6: 55439452-55439452 on Build GRCh38
rs895759849 Chr.6: 55439555-55439555 on Build GRCh38
rs900489223 Chr.6: 55439444-55439444 on Build GRCh38
rs761997706 Chr.6: 55439475-55439475 on Build GRCh38
rs764166829 Chr.6: 55439472-55439472 on Build GRCh38
rs368293693 Chr.6: 55439581-55439581 on Build GRCh38
rs376465811 Chr.6: 55439461-55439461 on Build GRCh38
rs772011384 Chr.6: 55439447-55439447 on Build GRCh38
rs534342292 Chr.6: 55439576-55439576 on Build GRCh38
rs141318879 Chr.6: 55439557-55439557 on Build GRCh38
rs760031668 Chr.6: 55439590-55439590 on Build GRCh38
rs778430958 Chr.6: 55439526-55439526 on Build GRCh38
rs750417611 Chr.6: 55439506-55439506 on Build GRCh38
rs758293303 Chr.6: 55439535-55439535 on Build GRCh38
rs547791788 Chr.6: 55439418-55439418 on Build GRCh38
rs756867993 Chr.6: 55439519-55439519 on Build GRCh38
rs766780507 Chr.6: 55439404-55439404 on Build GRCh38
rs936975951 Chr.6: 55439546-55439546 on Build GRCh38
rs750907226 Chr.6: 55439455-55439455 on Build GRCh38
rs569756227 Chr.6: 55439520-55439520 on Build GRCh38
rs751982134 Chr.6: 55439409-55439409 on Build GRCh38
rs780913445 Chr.6: 55439572-55439572 on Build GRCh38
rs767831751 Chr.6: 55439516-55439516 on Build GRCh38
rs911776834 Chr.6: 55439424-55439424 on Build GRCh38
rs539957076 Chr.6: 55439450-55439450 on Build GRCh38
rs550948890 Chr.6: 55439419-55439419 on Build GRCh38
rs768375500 Chr.6: 55439459-55439459 on Build GRCh38
rs746998535 Chr.6: 55439542-55439542 on Build GRCh38
rs773373401 Chr.6: 55439580-55439580 on Build GRCh38
rs570836545 Chr.6: 55439469-55439469 on Build GRCh38
rs760681895 Chr.6: 55439435-55439435 on Build GRCh38
rs756781335 Chr.6: 55439437-55439437 on Build GRCh38
rs774988492 Chr.6: 55439588-55439588 on Build GRCh38

View Full Product Details