Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 6387506-6387733 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTF2F1
Entrez Gene ID
2962
Gene Name
general transcription factor IIF subunit 1
Gene Aliases
BTF4, RAP74, TF2F1, TFIIF
UniGene
Hs.68257
Gene Chromosome Location
Chr.19: 6379569-6393280 on Build GRCh38
Transcripts
NM_002096.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6273352 c.363C>A p.N121K Chr.19: 6387523-6387523 on Build GRCh38
4082554 c.336C>T p.G112G Chr.19: 6387550-6387550 on Build GRCh38
6338751 c.372C>A p.Y124* Chr.19: 6387514-6387514 on Build GRCh38
1002879 c.353T>C p.V118A Chr.19: 6387533-6387533 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571387690 Chr.19: 6387573-6387573 on Build GRCh38
rs116536653 Chr.19: 6387547-6387547 on Build GRCh38
rs187840120 Chr.19: 6387611-6387611 on Build GRCh38
rs1004436109 Chr.19: 6387618-6387618 on Build GRCh38
rs753559231 Chr.19: 6387610-6387610 on Build GRCh38
rs949736255 Chr.19: 6387689-6387689 on Build GRCh38
rs563646482 Chr.19: 6387705-6387705 on Build GRCh38
rs879010523 Chr.19: 6387627-6387627 on Build GRCh38
rs759552767 Chr.19: 6387607-6387607 on Build GRCh38
rs770801416 Chr.19: 6387607-6387607 on Build GRCh38
rs755821462 Chr.19: 6387609-6387609 on Build GRCh38
rs759382070 Chr.19: 6387607-6387608 on Build GRCh38
rs200049909 Chr.19: 6387550-6387550 on Build GRCh38
rs370551251 Chr.19: 6387617-6387618 on Build GRCh38
rs970078607 Chr.19: 6387641-6387641 on Build GRCh38
rs967451560 Chr.19: 6387623-6387623 on Build GRCh38
rs141810642 Chr.19: 6387534-6387534 on Build GRCh38
rs150799496 Chr.19: 6387535-6387535 on Build GRCh38
rs116101441 Chr.19: 6387574-6387574 on Build GRCh38
rs777757680 Chr.19: 6387596-6387597 on Build GRCh38
rs757364526 Chr.19: 6387632-6387632 on Build GRCh38
rs748870523 Chr.19: 6387541-6387541 on Build GRCh38
rs533382464 Chr.19: 6387659-6387659 on Build GRCh38
rs958203117 Chr.19: 6387698-6387698 on Build GRCh38
rs1037622238 Chr.19: 6387618-6387618 on Build GRCh38
rs768926088 Chr.19: 6387579-6387579 on Build GRCh38
rs746408064 Chr.19: 6387551-6387551 on Build GRCh38
rs541198163 Chr.19: 6387640-6387640 on Build GRCh38
rs761814696 Chr.19: 6387592-6387592 on Build GRCh38
rs556998740 Chr.19: 6387600-6387600 on Build GRCh38
rs555842772 Chr.19: 6387613-6387613 on Build GRCh38
rs751407180 Chr.19: 6387605-6387605 on Build GRCh38
rs368991021 Chr.19: 6387612-6387612 on Build GRCh38
rs113534430 Chr.19: 6387608-6387608 on Build GRCh38
rs746748912 Chr.19: 6387606-6387607 on Build GRCh38
rs991768400 Chr.19: 6387653-6387653 on Build GRCh38
rs28580396 Chr.19: 6387649-6387649 on Build GRCh38
rs772283416 Chr.19: 6387569-6387569 on Build GRCh38
rs111231626 Chr.19: 6387624-6387624 on Build GRCh38
rs776689168 Chr.19: 6387589-6387589 on Build GRCh38
rs190348744 Chr.19: 6387677-6387677 on Build GRCh38

View Full Product Details