Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96327714-96327983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITPRIPL1
Entrez Gene ID
150771
Gene Name
inositol 1,4,5-trisphosphate receptor interacting protein-like 1
Gene Aliases
KIAA1754L
UniGene
Hs.65009
Gene Chromosome Location
Chr.2: 96325324-96330507 on Build GRCh38
Transcripts
NM_001008949.2 NM_001163523.1 NM_001163524.1 NM_001324490.1 NM_178495.5 XM_017003427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7158567 c.1153G>A p.D385N Chr.2: 96327784-96327784 on Build GRCh38
161816 c.1296G>T p.Q432H Chr.2: 96327903-96327903 on Build GRCh38
3426772 c.1114C>T p.R372* Chr.2: 96327745-96327745 on Build GRCh38
7158566 c.1177G>A p.D393N Chr.2: 96327784-96327784 on Build GRCh38
4164619 c.1334A>G p.H445R Chr.2: 96327941-96327941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568942457 Chr.2: 96327876-96327876 on Build GRCh38
rs773063556 Chr.2: 96327868-96327869 on Build GRCh38
rs371232545 Chr.2: 96327833-96327833 on Build GRCh38
rs766629569 Chr.2: 96327869-96327869 on Build GRCh38
rs148715117 Chr.2: 96327874-96327874 on Build GRCh38
rs201762466 Chr.2: 96327768-96327768 on Build GRCh38
rs778608412 Chr.2: 96327811-96327811 on Build GRCh38
rs748537189 Chr.2: 96327940-96327940 on Build GRCh38
rs764425130 Chr.2: 96327763-96327763 on Build GRCh38
rs748092469 Chr.2: 96327813-96327813 on Build GRCh38
rs754062415 Chr.2: 96327802-96327802 on Build GRCh38
rs951138746 Chr.2: 96327773-96327773 on Build GRCh38
rs539151763 Chr.2: 96327756-96327756 on Build GRCh38
rs756336241 Chr.2: 96327795-96327795 on Build GRCh38
rs777705215 Chr.2: 96327884-96327884 on Build GRCh38
rs745512936 Chr.2: 96327920-96327920 on Build GRCh38
rs774628223 Chr.2: 96327847-96327847 on Build GRCh38
rs745575790 Chr.2: 96327844-96327844 on Build GRCh38
rs777229457 Chr.2: 96327825-96327825 on Build GRCh38
rs750561164 Chr.2: 96327786-96327786 on Build GRCh38
rs769360860 Chr.2: 96327922-96327922 on Build GRCh38
rs772023375 Chr.2: 96327821-96327821 on Build GRCh38
rs772762201 Chr.2: 96327741-96327741 on Build GRCh38
rs890292332 Chr.2: 96327776-96327776 on Build GRCh38
rs746646034 Chr.2: 96327862-96327862 on Build GRCh38
rs533598116 Chr.2: 96327810-96327810 on Build GRCh38
rs752101843 Chr.2: 96327767-96327767 on Build GRCh38
rs758269052 Chr.2: 96327880-96327880 on Build GRCh38
rs772622332 Chr.2: 96327950-96327950 on Build GRCh38
rs748097785 Chr.2: 96327736-96327736 on Build GRCh38
rs756756215 Chr.2: 96327910-96327910 on Build GRCh38
rs75030364 Chr.2: 96327739-96327739 on Build GRCh38
rs754055738 Chr.2: 96327873-96327873 on Build GRCh38
rs770603089 Chr.2: 96327746-96327746 on Build GRCh38
rs780436364 Chr.2: 96327798-96327798 on Build GRCh38
rs768143282 Chr.2: 96327860-96327860 on Build GRCh38
rs557294833 Chr.2: 96327897-96327897 on Build GRCh38
rs1005098002 Chr.2: 96327826-96327826 on Build GRCh38
rs770516880 Chr.2: 96327838-96327838 on Build GRCh38
rs1015021102 Chr.2: 96327835-96327835 on Build GRCh38
rs982857512 Chr.2: 96327784-96327784 on Build GRCh38
rs749709553 Chr.2: 96327919-96327919 on Build GRCh38
rs768421706 Chr.2: 96327863-96327863 on Build GRCh38
rs141755021 Chr.2: 96327771-96327771 on Build GRCh38
rs193921120 Chr.2: 96327946-96327946 on Build GRCh38
rs35855657 Chr.2: 96327913-96327913 on Build GRCh38
rs537953486 Chr.2: 96327933-96327933 on Build GRCh38
rs527931884 Chr.2: 96327745-96327745 on Build GRCh38
rs747007175 Chr.2: 96327898-96327898 on Build GRCh38
rs775768030 Chr.2: 96327748-96327748 on Build GRCh38
rs769034930 Chr.2: 96327845-96327845 on Build GRCh38
rs114832056 Chr.2: 96327953-96327953 on Build GRCh38
rs571047143 Chr.2: 96327781-96327781 on Build GRCh38
rs779653246 Chr.2: 96327757-96327757 on Build GRCh38
rs769653732 Chr.2: 96327830-96327830 on Build GRCh38

View Full Product Details