Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 63609018-63609279 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HERC1
Entrez Gene ID
8925
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase family member 1
Gene Aliases
p532, p619
UniGene
Hs.210385
Gene Chromosome Location
Chr.15: 63608618-63833964 on Build GRCh38
Transcripts
NM_003922.3 XM_017022699.1 XM_017022700.1 XM_017022701.1 XM_017022702.1 XM_017022703.1 XM_017022704.1 XM_017022705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4690019 c.14481C>T p.A4827A Chr.15: 63609186-63609186 on Build GRCh38
434124 c.14417delC p.P4806fs*10 Chr.15: 63609250-63609250 on Build GRCh38
4690017 c.14544G>A p.S4848S Chr.15: 63609123-63609123 on Build GRCh38
1209556 c.14573A>G p.D4858G Chr.15: 63609094-63609094 on Build GRCh38
963694 c.14456C>T p.P4819L Chr.15: 63609211-63609211 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770034635 Chr.15: 63609119-63609119 on Build GRCh38
rs762677734 Chr.15: 63609060-63609061 on Build GRCh38
rs770912455 Chr.15: 63609182-63609182 on Build GRCh38
rs759762640 Chr.15: 63609147-63609147 on Build GRCh38
rs760564287 Chr.15: 63609163-63609163 on Build GRCh38
rs756534965 Chr.15: 63609211-63609211 on Build GRCh38
rs771024088 Chr.15: 63609132-63609132 on Build GRCh38
rs766318030 Chr.15: 63609118-63609118 on Build GRCh38
rs368675679 Chr.15: 63609210-63609210 on Build GRCh38
rs778065837 Chr.15: 63609213-63609213 on Build GRCh38
rs779806883 Chr.15: 63609186-63609186 on Build GRCh38
rs764852916 Chr.15: 63609176-63609176 on Build GRCh38
rs77087691 Chr.15: 63609057-63609057 on Build GRCh38
rs755853841 Chr.15: 63609072-63609072 on Build GRCh38
rs763763584 Chr.15: 63609168-63609168 on Build GRCh38
rs376035697 Chr.15: 63609079-63609079 on Build GRCh38
rs376844151 Chr.15: 63609142-63609142 on Build GRCh38
rs757210641 Chr.15: 63609175-63609175 on Build GRCh38
rs757834371 Chr.15: 63609185-63609185 on Build GRCh38
rs772019706 Chr.15: 63609240-63609240 on Build GRCh38
rs932306287 Chr.15: 63609058-63609058 on Build GRCh38
rs770900038 Chr.15: 63609054-63609054 on Build GRCh38
rs372164069 Chr.15: 63609144-63609144 on Build GRCh38
rs555703834 Chr.15: 63609065-63609065 on Build GRCh38
rs749757338 Chr.15: 63609124-63609124 on Build GRCh38
rs753855504 Chr.15: 63609172-63609172 on Build GRCh38
rs374249242 Chr.15: 63609181-63609181 on Build GRCh38
rs758066865 Chr.15: 63609114-63609114 on Build GRCh38
rs775558489 Chr.15: 63609155-63609155 on Build GRCh38
rs752473671 Chr.15: 63609071-63609071 on Build GRCh38
rs372178578 Chr.15: 63609237-63609237 on Build GRCh38
rs781414862 Chr.15: 63609115-63609115 on Build GRCh38
rs773811862 Chr.15: 63609123-63609123 on Build GRCh38
rs2228518 Chr.15: 63609108-63609108 on Build GRCh38
rs370346330 Chr.15: 63609096-63609096 on Build GRCh38
rs775817871 Chr.15: 63609246-63609246 on Build GRCh38
rs767843368 Chr.15: 63609153-63609153 on Build GRCh38
rs771365114 Chr.15: 63609228-63609228 on Build GRCh38
rs557894225 Chr.15: 63609215-63609215 on Build GRCh38
rs764146411 Chr.15: 63609078-63609078 on Build GRCh38
rs748603472 Chr.15: 63609116-63609116 on Build GRCh38

View Full Product Details