Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20770737-20770986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HP1BP3
Entrez Gene ID
50809
Gene Name
heterochromatin protein 1 binding protein 3
Gene Aliases
HP1-BP74, HP1BP74
UniGene
Hs.142442
Gene Chromosome Location
Chr.1: 20742677-20787809 on Build GRCh38
Transcripts
NM_016287.3 XM_005245875.4 XM_005245876.3 XM_005245877.4 XM_005245878.4 XM_005245879.4 XM_011541532.1 XM_011541533.1 XM_011541534.2 XM_017001393.1 XM_017001394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4438250 c.635A>G p.N212S Chr.1: 20770949-20770949 on Build GRCh38
4135949 c.654+2T>C p.? Chr.1: 20770928-20770928 on Build GRCh38
425235 c.653A>T p.Q218L Chr.1: 20770931-20770931 on Build GRCh38
4027843 c.618A>G p.A206A Chr.1: 20770966-20770966 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747651349 Chr.1: 20770882-20770882 on Build GRCh38
rs748367693 Chr.1: 20770907-20770909 on Build GRCh38
rs548418782 Chr.1: 20770776-20770776 on Build GRCh38
rs376382624 Chr.1: 20770772-20770772 on Build GRCh38
rs192899986 Chr.1: 20770900-20770900 on Build GRCh38
rs879843273 Chr.1: 20770838-20770839 on Build GRCh38
rs1015134382 Chr.1: 20770789-20770789 on Build GRCh38
rs756452835 Chr.1: 20770914-20770916 on Build GRCh38
rs938866456 Chr.1: 20770915-20770915 on Build GRCh38
rs142036561 Chr.1: 20770787-20770787 on Build GRCh38
rs528301990 Chr.1: 20770791-20770791 on Build GRCh38
rs755665206 Chr.1: 20770886-20770886 on Build GRCh38
rs189410202 Chr.1: 20770767-20770767 on Build GRCh38
rs779192038 Chr.1: 20770819-20770819 on Build GRCh38
rs914358513 Chr.1: 20770942-20770942 on Build GRCh38
rs985792660 Chr.1: 20770938-20770938 on Build GRCh38
rs745717743 Chr.1: 20770939-20770939 on Build GRCh38
rs774270602 Chr.1: 20770912-20770912 on Build GRCh38
rs777525689 Chr.1: 20770892-20770892 on Build GRCh38
rs781487202 Chr.1: 20770881-20770881 on Build GRCh38
rs989761789 Chr.1: 20770831-20770831 on Build GRCh38
rs201614762 Chr.1: 20770857-20770857 on Build GRCh38
rs770817851 Chr.1: 20770910-20770910 on Build GRCh38
rs748407592 Chr.1: 20770924-20770924 on Build GRCh38

View Full Product Details